Skip to Content
Merck
All Photos(1)

Key Documents

AR42J

93100618, rat pancreas, Pancreas cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

AR42J, 93100618

biological source

rat pancreas

growth mode

Semi-adherent aggregates

karyotype

Not specified

morphology

Pancreas cells

products

Amylase and exocrine enzymes

receptors

Gastrin/CCKB, Epidermal growth factor (EGF), Somatostatin

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Rat exocrine pancreatic tumour

Cell Line Description

Derived from a transplantable tumour of a rat exocrine pancreas. The line is tumourigenic in nude mice, and shows significant secretion of amylase and other exocrine enzymes.

Application

AR42J has been used to obtain a tumor mouse model by inoculating it subcutaneously in a female, athymic nude mouse. It has also been used to perform imaging studies.

Culture Medium

RPMI 1640 + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Cells grow in hollow spheroid colonies that can attach loosely. Split cultures 1:2 every 48 hours into fresh flasks, maintain cultures between 1-9x100,000 cells/ml. Adherent cells should be dislodged using 0.2% EDTA. N.B. High cell viability cannot be ex

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service