PANC-1 Cell Line human
87092802, human pancreas, Epithelial
Synonym(s):
PANC1 Cells, Panc-1-P Cells
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41106514
eCl@ss:
32190102
Recommended Products
Product Name
PANC-1 Cell Line human, 87092802, from human pancreas
biological source
human pancreas
growth mode
Adherent
karyotype
2n = 46, hypertriploid
morphology
Epithelial
products
Not specified
receptors
Not specified
technique(s)
cell culture | mammalian: suitable
relevant disease(s)
cancer
shipped in
dry ice
storage temp.
−196°C
Cell Line Origin
Human Caucasian pancreas
Cell Line Description
Established from a pancreatic carcinoma of ductal origin from a 56-year-old Caucasian male. Cells possess the type B phenotype for G6PD. The Y chromosome could not be detected in this cell line by short tandem repeat (STR)-PCR analysis when tested at ECACC. It is a known phenomenon that due to the increased genetic instability of cancer cell lines the Y chromosome can be rearranged or lost resulting in lack of detection. The cell line is identical to the source provided by the depositor based on the STR-PCR analysis.
Application
Human PANC-1 cell line has been used to study the role of the intermediate filament keratin reorganization on the alignment and migration of cancer cells. It has also been used to study calcium-mediated actin reset (CaAR) in response to physiological changes.
DNA Profile
STR-PCR Data: Amelogenin: X
CSF1PO: 10,12
D13S317: 11
D16S539: 11
D5S818: 11,13
D7S820: 8,10
THO1: 7,8
TPOX: 8,11
vWA: 15
CSF1PO: 10,12
D13S317: 11
D16S539: 11
D5S818: 11,13
D7S820: 8,10
THO1: 7,8
TPOX: 8,11
vWA: 15
Culture Medium
DMEM + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).
Subculture Routine
Split sub-confluent cultures (70-80%) 1:3 to 1:6 i.e. seeding at 2-4x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C.
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service