Skip to Content
Merck
All Photos(1)

Key Documents

SAOS-2 Cell Line human

NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA, 89050205, human bone, Epithelial-like

Synonym(s):

SAOS2 Cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

biological source

human bone

packaging

tube of 5 μg 89050205-DNA-5UG
pkg of vial of cells 89050205-1VL

growth mode

Adherent

karyotype

2n = 46, (P1) Hyperploid to hypopentaploid

morphology

Epithelial-like

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human primary osteogenic sarcoma

Cell Line Description

Reported to have been derived from an 11 year old female Caucasian. The patient was treated with RTG, methotrexate, adriamycin, vincristine, cytoxan, and aramycin-C. HLA cell line phenotype: A2,3;Bw16,w47.

Application

SAOS-2 cell line has been used to study the importance of cyclin D1 for the activity of lithocholic acid hydroxyamide (LCAHA). It has also been used to study the effect of chromium-enriched diamond-like carbon (DLC) layers on cell adhesion and proliferation.

DNA Profile

STR-PCR Data: Amelogenin: X
CSF1PO: 10
D13S317: 12,13
D16S539: 12,13
D5S818: 12
D7S820: 8,10
THO1: 6,9
TPOX: 8
vWA: 18

Culture Medium

McCoy′s 5a + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:6 i.e. seeding at 2-4x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service