Skip to Content
Merck
All Photos(1)

Key Documents

SW480 Cell Line human

87092801, human colon, Epithelial

Synonym(s):

SW-480 Cells, SW480E Cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

SW480 Cell Line human, 87092801

biological source

human colon

description

Human colon adenocarcinoma

growth mode

Adherent

karyotype

Not specified

morphology

Epithelial

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human colon adenocarcinoma

Cell Line Description

Derived from a grade 3-4 colon adenocarcinoma. The initial cultures contained a mixture of epithelial and bipolar cells, but subsequently epithelial cells predominated. The cells produce CEA.

Application

Human SW480 cell line has been used to study the antiproliferative effects of F. mughlea Pesmen on cancer cells. It has also been used as an in vitro model to study key cellular processes required for metastasis. The cell line is used to study the effects of the transforming growth factor-β (TGF-β) pathway and heat shock protein 90 (Hsp90) on the growth of cells.

DNA Profile

STR-PCR Data: Amelogenin: X
CSF1PO: 13,14
D13S317: 12
D16S539: 13
D5S818: 13
D7S820: 8
THO1: 8
TPOX: 11
vWA: 16

Culture Medium

L15 + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:2 to 1:8 i.e. seeding at 2-3x10,000 cells/cm2 using 0.25% trypsin; 37°C. Medium change every 4-5 days. CO2 is not required.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service