Skip to Content
Merck
All Photos(1)

Documents

HCT-15

91030712, human colon, Epithelial-like

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

HCT-15, 91030712

biological source

human colon

growth mode

Adherent

karyotype

2n = 46, quasidiploid

morphology

Epithelial-like

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human colon adenocarcinoma

Cell Line Description

Colorectal adenocarcinoma which is tumorigenic in nude mice.The four cell lines: DLD-1 (90102540), HCT-15 (91030712), HCT-8 (90032006) and HRT-18 (86040306) have been shown to have a common genetic origin.

Application

HCT-15 cell line has been used to study the role and efficiency of nexrutine (NX), an extract from Phellodendron amurense, against colon cancer.

Culture Medium

RPMI 1640 + 2mM Glutamine + 20% Foetal Bovine Serum (FBS) (heat inactivated).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:6 i.e. seeding at 2-4x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service