Skip to Content
Merck
All Photos(1)

Documents

BUA462-EP

Buckets for Eppendorf® A-4-62 Rotor

aerosol-tight capable, capacity 250 mL, pack of 4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103910

feature

aerosol-tight capable

packaging

pack of 4

manufacturer/tradename

Eppendorf® 022638084 (NA)
Eppendorf® 5810716004 (EU)

technique(s)

centrifugation: suitable

capacity

250 mL

compatibility

for use with Rotor A-4-62

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

  • Rectangular buckets have a maximum speed of 3,220 × g (4,000 rpm)
  • Compatible with A-4-62 swing bucket rotor for 5810/5810R centrifuge
  • Autoclavable at 121 °C for 20 min

Legal Information

Eppendorf is a registered trademark of Eppendorf AG

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service