Skip to Content
Merck
All Photos(1)

Documents

EP022638033

Caps for Eppendorf® A-4-62 Rotor

aerosol-tight, for 5804/5804R & 5810/5810R, for 250 mL rectangular buckets

Synonym(s):

Lid for Eppendorf® FA-45-48-11 Rotor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103913

feature

aerosol-tight
for 250 mL rectangular buckets

packaging

pack of 2

manufacturer/tradename

Eppendorf® 022638033 (NA)

technique(s)

centrifugation: suitable

compatibility

for use with 5804 R
for use with 5804
for use with 5810 R
for use with 5810

Related Categories

Legal Information

Eppendorf is a registered trademark of Eppendorf AG

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service