Skip to Content
Merck
All Photos(1)

Key Documents

EP022638068

Buckets for Eppendorf® A-4-62 Rotor

aerosol-tight capable, (4 MTP or 1 DWP), pack of 4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103910

feature

aerosol-tight capable

packaging

pack of 4

manufacturer/tradename

Eppendorf® 022638068 (NA)

technique(s)

centrifugation: suitable

capacity

(4 MTP or 1 DWP)

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

  • Plate buckets have a maximum speed of 2,750 × g (4,000 rpm)
  • Each bucket accommodates 4 microtiter plates (MTP) or 1 deep well plate (DWP)
  • Compatible with A-4-62 MTP swing bucket rotor for 5810/5810R centrifuge
  • Autoclavable at 121 °C for 20 min

Legal Information

Eppendorf is a registered trademark of Eppendorf AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service