Skip to Content
Merck
All Photos(1)

Key Documents

A165

Sigma-Aldrich

Rp-Adenosine 3′,5′-cyclic monophosphorothioate triethylammonium salt

powder, ≥98% (HPLC)

Synonym(s):

Rp-Cyclic 3′,5′-hydrogen phosphorothioate adenosine triethylammonium salt, Rp-cAMPS triethylammonium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H11N5O5PS · C6H16N
CAS Number:
Molecular Weight:
446.46
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

H2O: 10 mg/mL

λmax

258 nm

storage temp.

−20°C

SMILES string

CCN(CC)CC.Nc1ncnc2n(cnc12)[C@@H]3O[C@@H]4COP(O)(=S)O[C@H]4[C@H]3O

InChI

1S/C10H12N5O5PS.C6H15N/c11-8-5-9(13-2-12-8)15(3-14-5)10-6(16)7-4(19-10)1-18-21(17,22)20-7;1-4-7(5-2)6-3/h2-4,6-7,10,16H,1H2,(H,17,22)(H2,11,12,13);4-6H2,1-3H3/t4-,6-,7-,10-,21?;/m1./s1

InChI key

OXIPZMKSNMRTIV-NVGWRVNNSA-N

Related Categories

Application

Rp-Adenosine 3′,5′-cyclic monophosphorothioate triethylammonium salt has been used as an antagonist of cAMP to block the cAMP-PKA signal pathway.

Biochem/physiol Actions

Rp-Adenosine 3′,5′-cyclic monophosphorothioate triethylammonium salt is capable of inhibiting protein kinase A (PKA).
Rp-Diastereomer of adenosine-3′,5′-cyclic monophosphothioate. Specific membrane-permeable inhibitor of activation by cAMP of cAMP-dependent protein kinase I and II; resistant toward cyclic nucleotide phosphodiesterases; blocks cAMP-mediated effects in numerous systems.

Features and Benefits

This compound is a featured product for Cyclic Nucleotide research. Click here to discover more featured Cyclic Nucleotide products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Caution

Extremely hygroscopic. Store desiccated.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Takashi Matsuura et al.
Journal of oral science, 60(4), 634-637 (2018-06-01)
Once a tooth develops deep caries and the dental pulp tissue is irreversibly infected, the infected dental pulp tissue should be removed, and filling material should be placed in the root canal. Endodontically treated teeth are prone to root fracture
Role of PLC-PIP2 and cAMP-PKA signal pathways in radiation-induced immune-suppressing effect
Dong JC, et al.
Biomedical and Environmental Sciences : Bes, 27(1), 27-34 (2014)
Carmen Müllner et al.
The FEBS journal, 276(21), 6218-6226 (2009-09-22)
Besides being activated by G-protein beta/gamma subunits, G-protein activated potassium channels (GIRKs) are regulated by cAMP-dependent protein kinase. Back-phosphorylation experiments have revealed that the GIRK1 subunit is phosphorylated in vivo upon protein kinase A activation in Xenopus oocytes, whereas phosphorylation
Shaomin Peng et al.
Investigative ophthalmology & visual science, 51(6), 3216-3225 (2010-01-01)
Bevacizumab and ranibizumab are currently used to treat age-related macular degeneration by neutralizing vascular endothelial growth factor (VEGF). In this study, the potential side effects on the outer blood-retinal barrier were examined. Human fetal RPE (hfRPE) cells were used because
PKA-independent cAMP stimulation of white adipocyte exocytosis and adipokine secretion: modulations by Ca2+ and ATP
Komai AM, et al.
The Journal of Physiology, 592(23), 5169-5186 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service