Skip to Content
Merck
All Photos(1)

Key Documents

A7410

Sigma-Aldrich

Amiloride hydrochloride hydrate

≥98% (HPLC), powder, T-type calcium channel blocker

Synonym(s):

N-Amidino-3,5-diamino-6-chloropyrazinecarboxamide hydrochloride hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H8ClN7O · HCl · xH2O
CAS Number:
Molecular Weight:
266.09 (anhydrous basis)
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Product Name

Amiloride hydrochloride hydrate, ≥98% (HPLC), powder

Quality Level

Assay

≥98% (HPLC)

form

powder

color

yellow

mp

285-288 °C (dec.)

solubility

H2O: 50 mg/mL, clear, yellow-green

originator

Perrigo

storage temp.

room temp

SMILES string

O.Cl.NC(=N)NC(=O)c1nc(Cl)c(N)nc1N

InChI

1S/C6H8ClN7O.ClH.H2O/c7-2-4(9)13-3(8)1(12-2)5(15)14-6(10)11;;/h(H4,8,9,13)(H4,10,11,14,15);1H;1H2

InChI key

WDZJJRLYFQNCQL-UHFFFAOYSA-N

Application

Amiloride is a selective T-type calcium channel blocker, an epithelial sodium channel blocker and a selective inhibitor of urokinase plasminogen activator (uPA). Amiloride has been used in a study to develop an alternative treatment for constipation.

Biochem/physiol Actions

Selective T-type Ca2+ and epithelial sodium channel blocker
Selective T-type calcium channel blocker and blocker of epithelial sodium channel. Selective inhibitor of urokinase plasminogen activator (uPA).

Features and Benefits

This compound is a featured product for ADME Tox research. Click here to discover more featured ADME Tox products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound is featured on the Acid-Sensing (Proton-gated) Ion Channels (ASICs) and Imidazoline Binding Sites pages of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
This compound was developed by Perrigo. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Caution

Protect from light.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tengis S Pavlov et al.
Journal of the American Society of Nephrology : JASN, 24(7), 1053-1062 (2013-04-20)
Various stimuli, including hormones and growth factors, modulate epithelial sodium channels (ENaCs), which fine-tune Na(+) absorption in the kidney. Members of the EGF family are important for maintaining transepithelial Na(+) transport, but whether EGF influences ENaC, perhaps mediating salt-sensitive hypertension
Tiphaine Dejouvencel et al.
Blood, 115(10), 2048-2056 (2009-12-10)
Fibrinolysis and pericellular proteolysis depend on molecular coassembly of plasminogen and its activator on cell, fibrin, or matrix surfaces. We report here the existence of a fibrinolytic cross-talk mechanism bypassing the requirement for their molecular coassembly on the same surface.
Cristina Matthewman et al.
American journal of physiology. Cell physiology, 311(6), C920-C930 (2016-10-21)
Hyperactivated DEG/ENaC channels cause neuronal death mediated by intracellular Ca2+ overload. Mammalian ASIC1a channels and MEC-4(d) neurotoxic channels in Caenorhabditis elegans both conduct Na+ and Ca2+, raising the possibility that direct Ca2+ influx through these channels contributes to intracellular Ca2+
Stephanie Denk et al.
Journal of immunology (Baltimore, Md. : 1950), 198(12), 4846-4854 (2017-05-12)
During sepsis, excessive activation of the complement system with generation of the anaphylatoxin C5a results in profound disturbances in crucial neutrophil functions. Moreover, because neutrophil activity is highly dependent on intracellular pH (pH
Hiu-Fung Yuen et al.
PloS one, 8(1), e54211-e54211 (2013-02-02)
The Hippo pathway restricts the activity of transcriptional coactivators TAZ (WWTR1) and YAP. TAZ and YAP are reported to be overexpressed in various cancers, however, their prognostic significance in colorectal cancers remains unstudied. The expression levels of TAZ and YAP

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service