Skip to Content
Merck
All Photos(1)

Key Documents

275R-3

Sigma-Aldrich

Ki-67 (MRQ-64) Rabbit Monoclonal Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

rabbit

Quality Level

100
500

conjugate

unconjugated

antibody form

culture supernatant

antibody product type

primary antibodies

clone

MRQ-64, monoclonal

description

For In Vitro Diagnostic Use in Select Regions

form

buffered aqueous solution

species reactivity

human

packaging

vial of 0.1 mL concentrate (275R-34)
vial of 0.1 mL concentrate Research Use Only (275R-34-RUO)
vial of 0.5 mL concentrate (275R-35)
vial of 1.0 mL concentrate (275R-36)
vial of 1.0 mL concentrate Research Use Only (275R-36-RUO)
vial of 1.0 mL pre-dilute Research Use Only (275R-37-RUO)
vial of 1.0 mL pre-dilute ready-to-use (275R-37)
vial of 25.0 mL pre-dilute ready-to-use (275R-30)
vial of 25.0 mL pre-dilute ready-to-use Research Use Only (275R-30-RUO)
vial of 7.0 mL pre-dilute ready-to-use (275R-38)
vial of 7.0 mL pre-dilute ready-to-use Research Use Only (275R-38-RUO)

manufacturer/tradename

Cell Marque

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:25-1:100 (concentrated)

isotype

IgG

control

Burkitt lymphoma, breast (ductl carcinoma in situ), tonsil

shipped in

wet ice

storage temp.

2-8°C

visualization

nuclear

Gene Information

human ... MKI67(4288)

General description

Ki-67 (MRQ-64) is an antibody designed to target Ki-67 antigen-a nuclear protein encoded by the MKI67 gene whose presence correlates with G1, S, G2, and the mitotic phases of the cell cycle.1 Though its specific role is largely postulated and still yet to be determined, Ki-67′s presence during cellular division makes it an excellent target for labeling rapidly growing malignancies.2 Given that its presence correlates with cell proliferation, the immunohistochemical labeling of Ki-67 antigen may be useful in characterizing a lesion′s growth fraction. It has been demonstrated that identifying this antigen by IHC is useful in characterizing lesions of the breast, lymphoid cells, astrocytes, and uterus.3-9

Quality

United States - IVD
Canada - IVD
European Union - IVD
Japan - RUO

Linkage

Ki-67 Positive Control Slides, Product No. 275S, are available for immunohistochemistry (formalin-fixed, paraffin-embedded sections).

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide.

Preparation Note

Download the IFU specific to your product lot and format
Note: This requires a keycode which can be found on your packaging or product label.

Download the latest released IFU
Note: This IFU may not apply to your specific product lot.

Other Notes

For Technical Service please contact: 800-665-7284 or email: service@cellmarque.com

Legal Information

Cell Marque is a trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shih-Sung Chuang et al.
American journal of clinical pathology, 128(4), 558-564 (2007-09-19)
Burkitt lymphoma (BL) is characterized by c-myc translocation and CD10+/bc-2-/bcl-6+ with a very high Ki-67 proliferation index (PI). Occasional diffuse large B-cell lymphomas may exhibit a very high PI with or without a starry-sky pattern (DLBCL-HPSS). We compared 28 consecutive
Klaus Mayerhofer et al.
Acta obstetricia et gynecologica Scandinavica, 83(11), 1085-1088 (2004-10-19)
The aim of the current study was to evaluate the expression of Ki-67 in uterine smooth muscle tumors, comparing leiomyomas, uterine smooth muscle tumors of uncertain malignant potential (STUMP) and uterine leiomyosarcomas (LMS) and to prove the accuracy of a
Nandish Vastrad Shivaprasad et al.
Journal of neurosciences in rural practice, 7(4), 510-514 (2016-10-04)
Astrocytomas are the most common primary tumor of the central nervous system. The distinction between different tumor grades can be tested despite criteria given by the World Health Organization (WHO). Ki-67 is a potent biological marker used in grading of
Xiaoming Sun et al.
Chromosoma, 127(2), 175-186 (2018-01-13)
Ki-67 protein has been widely used as a proliferation marker for human tumor cells for decades. In recent studies, multiple molecular functions of this large protein have become better understood. Ki-67 has roles in both interphase and mitotic cells, and
Adi Broyde et al.
American journal of hematology, 84(6), 338-343 (2009-04-23)
Expression of Ki-67, a nuclear antigen protein present in all cycling cells, is used to determine the growth fraction of tumors. The aim of this study was to evaluate the role and prognostic significance of the Ki-67 proliferation index (PI)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service