Skip to Content
Merck
All Photos(1)

Key Documents

103R-9

Sigma-Aldrich

CD3 (MRQ-39) Rabbit Monoclonal Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

100
500

conjugate

unconjugated

antibody form

culture supernatant

antibody product type

primary antibodies

clone

MRQ-39, monoclonal

description

For In Vitro Diagnostic Use in Select Regions (See Chart)

form

buffered aqueous solution

species reactivity

human

packaging

pkg of 0.1 mL concentrate (103r-94)
pkg of 0.5 mL concentrate (103R-95)
pkg of 1.0 mL concentrate (103R-96)
pkg of 1.0 mL predilute (103R-97)
pkg of 7.0 mL predilute (103R-98)

manufacturer/tradename

Cell Marque®

IVD

for in vitro diagnostic use

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:100-1:500 (concentrated)

isotype

IgG1

control

tonsil

shipped in

wet ice

storage temp.

2-8°C

visualization

membranous

Gene Information

human ... CD3E(916)

General description

Anti-CD3 has been considered the best all around T-cell marker. This antibody reacts with an antigen present in early thymocytes. The positive staining of this marker may represent a sign of early commitment to the T-Cell lineage.

Quality


IVD

IVD

IVD

RUO

Linkage

CD3 Positive Control Slides, Product No. 103S, are available for immunohistochemistry (formalin-fixed, paraffin-embedded sections).

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide

Preparation Note

Download the IFU specific to your product lot and formatNote: This requires a keycode which can be found on your packaging or product label.

Other Notes

For Technical Service please contact: 800-665-7284 or email: [email protected]

Legal Information

Cell Marque is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E A Clark et al.
Immunology today, 10(7), 225-228 (1989-07-01)
During 1987, striking advances were made in defining the receptors and ligands for cell-to-cell adhesion interactions involving leukocytes. In 1988, two major leukocyte differentiation antigens, CD10 (cALLA) and CD45 (LCA, T200), were shown to be enzymes while two other markers
SM Denning, et al.
Leucocyte Typing III, 144-147 (1987)
Kennosuke Karube et al.
The American journal of surgical pathology, 27(10), 1366-1374 (2003-09-26)
We studied the morphologic, immunohistochemical, and clinical characteristics of 158 cases of lymphoblastic lymphoma. Based on immunophenotyping and cell lineage, cases were classified into B-cell type (CD20,CD19 or CD79a+, n = 53), T-cell type (surface CD3+, n = 84), and
P C Beverley et al.
European journal of immunology, 11(4), 329-334 (1981-04-01)
The properties of human lymphocyte fractions isolated either by sheep red cell(E) rosetting or by fluorescence-activated cell sorting after staining with UCHT1 monoclonal anti-T cell antibody have been compared. Two populations of E+ cells with very different phenotype and function
D Campana et al.
Journal of immunology (Baltimore, Md. : 1950), 138(2), 648-655 (1987-01-15)
Anti-CD3 (T3) Ab reacting with different proportions of thymocytes (anti-CD3a: UCHT1, anti-CD3b: T10B9, and anti-CD3c: OKT3) were tested for cytoplasmic (cCD3) and membrane (mCD3) expression in the bone marrow, thymus, and blood in man and selected primates. The expression of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service