Skip to Content
Merck
All Photos(1)

Key Documents

24368C

SAFC

EX-CELL® Advanced CHO Feed 1 (without glucose)

Pharma Manufacturing

Synonym(s):

EX-CELL® Advanced CHO Feed 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.71

product line

EX-CELL®

Quality Level

form

dry powder

technique(s)

cell culture | mammalian: suitable (suspension)

shipped in

ambient

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

EX-CELL® Advanced CHO Feed 1 (without glucose) is chemically-defined and contains no animal derived components. This feed is intended to be a part of a fed-batch platform, and used with EX-CELL® Advanced CHO Fed-batch Medium (14366C or 24366C). This newest platform from SAFC has been specifically developed to support growth and productivity across a diverse set of Chinese Hamster Ovary (CHO) cells in fed-batch cultures while still allowing for flexibility in the adjustment of protein quality attributes. This contemporary fed-batch platform was developed using a strategy of Multivariate Analysis to establish correlations in which the formulation components influence critical process and product attributes such as titer, fold expansion, receptor expression, and glycosylation. This platform contains no hypoxanthine and thymidine, L-glutamine, phenol red or 2-mercaptoethanol (2-ME).

Linkage

Submit the EX-CELL Advanced CHO Fed-batch request form to get in touch with a SAFC representative.

Quantity

Formulated to contain 34.1 grams of powder per liter of medium.

Legal Information

EX-CELL is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pao-Chun Lin et al.
mAbs, 11(5), 965-976 (2019-05-03)
Chinese hamster ovary (CHO) cells are the biopharmaceutical industry's primary means of manufacturing therapeutic proteins, including monoclonal antibodies. The major challenge in cell line development for the production of recombinant biopharmaceuticals lies in generating and isolating rare high-producing stable clones

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service