Skip to Content
Merck
All Photos(1)

Key Documents

14361C

SAFC

EX-CELL® CD CHO Serum-Free Medium for CHO Cells, Chemically Defined

Animal-component free, with hypoxanthine, with thymidine, without L-glutamine, liquid, sterile-filtered, suitable for cell culture

Synonym(s):

EX-CELL® CD CHO Serum-Free Medium for CHO Cells, Chemically Defined

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352207
NACRES:
NA.75

description

for research or for further manufacturing use

sterility

sterile-filtered

form

liquid

technique(s)

cell culture | mammalian: suitable

components

NaHCO3: 2.10 g/L
phenol red: no
HEPES: no
glucose: 5.6 g/L

shipped in

ambient

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

EX-CELL CD CHO is a chemically defined, animal-component free, serum-free medium developed for the long-term growth of Chinese Hamster Ovary (CHO) cells and expression of antibodies or protein products in suspension cell culture. CHO suspension cultures can be subcultured directly into EX-CELL CD CHO from serum-supplemented or serum-free medium with little or no adaptation. EX-CELL CD CHO (Catalog No. 14361C) is formulated with hypoxanthine and thymidine, making it an appropriate medium for applications that do not require the selective pressure of a hypoxanthine/thymidine (HT)-deficient medium, such as CHO-K1 and Glutamine Synthetase (GS System) users.

Legal Information

EX-CELL is a registered trademark of Merck KGaA, Darmstadt, Germany
GS System is a trademark of Lonza Group Ltd.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andrew S Flies et al.
Science advances, 6(27) (2020-09-17)
Around 40% of humans and Tasmanian devils (Sarcophilus harrisii) develop cancer in their lifetime, compared to less than 10% for most species. In addition, devils are affected by two of the three known transmissible cancers in mammals. Immune checkpoint immunotherapy

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service