Skip to Content
Merck
All Photos(1)

Key Documents

76871

Sigma-Aldrich

Pentane

puriss. p.a., ≥99.0% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3CH3
CAS Number:
Molecular Weight:
72.15
Beilstein:
969132
EC Number:
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.06
technique(s):
GC/GC: suitable
bp:
35-36 °C (lit.)
vapor pressure:
26.98 psi

vapor density

2.48 (vs air)

Quality Level

vapor pressure

26.98 psi

grade

puriss. p.a.

Assay

≥99.0% (GC)

form

liquid

autoignition temp.

500 °F

expl. lim.

~8.3 %

technique(s)

GC/GC: suitable

impurities

≤0.05% water

evapn. residue

≤0.002%

refractive index

n20/D 1.358 (lit.)
n20/D 1.358

bp

35-36 °C (lit.)

mp

−130 °C (lit.)

solubility

ethanol: soluble(lit.)

density

0.626 g/mL at 25 °C (lit.)

cation traces

Al: ≤0.5 mg/kg
Ba: ≤0.1 mg/kg
Bi: ≤0.1 mg/kg
Ca: ≤0.5 mg/kg
Cd: ≤0.05 mg/kg
Co: ≤0.02 mg/kg
Cr: ≤0.02 mg/kg
Cu: ≤0.02 mg/kg
Fe: ≤0.1 mg/kg
K: ≤0.5 mg/kg
Li: ≤0.1 mg/kg
Mg: ≤0.1 mg/kg
Mn: ≤0.02 mg/kg
Mo: ≤0.1 mg/kg
Na: ≤0.5 mg/kg
Ni: ≤0.02 mg/kg
Pb: ≤0.1 mg/kg
Sr: ≤0.1 mg/kg
Zn: ≤0.1 mg/kg

SMILES string

CCCCC

InChI

1S/C5H12/c1-3-5-4-2/h3-5H2,1-2H3

InChI key

OFBQJSOFQDEBGM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pentane (n-pentane), a hydrocarbon, is a colorless liquid having 3 structural isomeric forms. It is a promising solvent for the dissolution of oils, fats and resins. Its hydroisomerization using a nanocrystalline beta (BEA) zeolite has been proposed. It has been reported as one of the products formed from the peroxidation of unsaturated fatty acid. It isomerizes to iso-pentane in the presence of ZSM-5 (Zeolite Socony Mobil-5) zeolite catalysts under hydrogen and nitrogen atmosphere.

Application

Pentane may be used for the fabrication of planar lipid bilayers on the films of amorphous PTFE (Polytetrafluoroethylene) by “painting technique”.

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

-40.0 °F

Flash Point(C)

-40 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hydroisomerization of n-pentane over hybrid catalysts containing a supported hydrogenation catalyst.
Fujimoto K, et al.
Applied Catalysis A: General, 91(2), 81-86 (1992)
Eagleson M.
Concise Encyclopedia Chemistry, 783-783 (1994)
Anna Svensson et al.
Carbohydrate polymers, 92(1), 775-783 (2012-12-12)
Dry ultra-porous cellulose fibres were obtained using a liquid exchange procedure in which water was replaced in the following order: water, methanol, acetone, and finally pentane; thereafter, the fibres were dried with Ar(g). The dry samples (of TEMPO-oxidized dissolving pulp)
Keisuke Maruyama et al.
Bioorganic & medicinal chemistry letters, 20(22), 6661-6666 (2010-10-05)
We have proposed a multi-template approach for drug development, focusing on similar fold structures of proteins, and have effectively generated lead compounds for several drug targets. Modification of these polypharmacological lead compounds is then needed to generate target-selective compounds. In
Assessment of lipid peroxidation in newborn infants and rabbits by measurements of expired ethane and pentane: influence of parenteral lipid infusion.
Wispe JR, et al.
Pediatric Research, 19(4), 374-379 (1985)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service