Skip to Content
Merck
All Photos(2)

Key Documents

1.00882

Supelco

n-Pentane

for gas chromatography ECD and FID SupraSolv®

Synonym(s):

Pentane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3CH3
CAS Number:
Molecular Weight:
72.15
Beilstein:
969132
MDL number:
UNSPSC Code:
41116105
EC Index Number:
203-692-4
NACRES:
NA.03
Assay:
≥99.8% (GC)
technique(s):
gas chromatography (GC): suitable
bp:
35-36 °C (lit.)
Pricing and availability is not currently available.

vapor density

2.48 (vs air)

Quality Level

product line

SupraSolv®

Assay

≥99.8% (GC)

form

liquid

autoignition temp.

500 °F

potency

>2000 mg/kg LD50, oral (Rat)

expl. lim.

~8.3 %

technique(s)

gas chromatography (GC): suitable

refractive index

n20/D 1.358 (lit.)

bp

35-36 °C (lit.)

mp

−130 °C (lit.)

transition temp

flash point -48 °C (refers to pure substance)

solubility

0.4 g/L

density

0.626 g/mL at 25 °C (lit.)

storage temp.

2-30°C

SMILES string

CCCCC

InChI

1S/C5H12/c1-3-5-4-2/h3-5H2,1-2H3

InChI key

OFBQJSOFQDEBGM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

SupraSolv® ECD and FID is specially developed for gas chromatography in combination with ECD (Electron Capture Detector) and FID (Flame Ionization detector). SupraSolv® solvents offer the largest specified retention time range, a clear baseline and a minimum signal-to-noise ratio leading to reliable and reproducible analysis results.
Recovered from special distillation cuts, they are tested for specific detectors to ensure high application security.

Application

Ideal for GC applications including sensitive detection in residue and environmental analysis .

Other Notes

Explore various lab safety accessories and equipment for safe handling of solvents to increase your safety level from the first usage.

Legal Information

SUPRASOLV is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

-40.0 °F

Flash Point(C)

-40 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service