Skip to Content
Merck
All Photos(1)

Key Documents

60489

Sigma-Aldrich

Pentane

≥99% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3CH3
CAS Number:
Molecular Weight:
72.15
Beilstein:
969132
EC Number:
MDL number:
UNSPSC Code:
12191502
PubChem Substance ID:
NACRES:
NA.21
Assay:
≥99% (GC)
bp:
35-36 °C (lit.)
vapor pressure:
26.98 psi

vapor density

2.48 (vs air)

Quality Level

vapor pressure

26.98 psi

Assay

≥99% (GC)

form

liquid

autoignition temp.

500 °F

expl. lim.

~8.3 %

refractive index

n20/D 1.358 (lit.)

bp

35-36 °C (lit.)

mp

−130 °C (lit.)

density

0.626 g/mL at 25 °C (lit.)

SMILES string

CCCCC

InChI

1S/C5H12/c1-3-5-4-2/h3-5H2,1-2H3

InChI key

OFBQJSOFQDEBGM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Pentane (n-pentane) is a hydrocarbon. Its hydroisomerization using a nanocrystalline beta (BEA) zeolite has been proposed. It has been reported as one of the products formed from the peroxidation of unsaturated fatty acid.

Application


  • Catalytic Cracking to Light Olefins: Pentane is extensively used in studies focused on catalytic cracking processes. Recent research has explored the use of extra-framework aluminum in ZSM-5 zeolites to enhance the production of light olefins from pentane. This application is crucial for improving the efficiency of catalytic processes and the production of valuable petrochemical feedstocks (He et al., Journal of Analytical and Applied Pyrolysis, 2022).

Other Notes

The article number 60489-6X1L-M will be discontinued. Please order the single bottle 60489-1L-M which is physically identical with the same exact specifications.
The article number 60489-6X1L-R will be discontinued. Please order the single bottle 60489-1L-R which is physically identical with the same exact specifications.
Pertains only in Germany: unversteuert

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

-40.0 °F

Flash Point(C)

-40 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

James Clarke et al.
Nature nanotechnology, 4(4), 265-270 (2009-04-08)
A single-molecule method for sequencing DNA that does not require fluorescent labelling could reduce costs and increase sequencing speeds. An exonuclease enzyme might be used to cleave individual nucleotide molecules from the DNA, and when coupled to an appropriate detection
Assessment of lipid peroxidation in newborn infants and rabbits by measurements of expired ethane and pentane: influence of parenteral lipid infusion.
Wispe JR, et al.
Pediatric Research, 19(4), 374-379 (1985)
Hydroisomerization of pentane, hexane, and heptane for improving the octane number of gasoline.
Chica A and Corma A.
J. Catal., 187(1), 167-176 (1999)
Fuming Chen et al.
Langmuir : the ACS journal of surfaces and colloids, 27(15), 9174-9181 (2011-07-01)
Graphene oxide (GO) can be viewed as an amphiphilic soft material, which form thin films at organic solvent-water interfaces. However, organic solvent evaporation provides little driving force, which results in slow GO transfer in aqueous phase, thus dawdling GO film
Michael Polet et al.
Biomedical chromatography : BMC, 27(2), 259-266 (2012-07-21)
The administration of anabolic steroids is one of the most important issues in doping control and is detectable through a change in the carbon isotopic composition of testosterone and/or its metabolites. Gas chromatography-combustion-isotope ratio mass spectrometry (GC-C-IRMS), however, remains a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service