Skip to Content
Merck
All Photos(2)

Key Documents

563935

Sigma-Aldrich

2-Propanol

70% in H2O

Synonym(s):

Isopropanol, 2-Propanol, 2-Propanol solution, IPA, Isopropyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.05
bp:
80.9-83.2 °C
vapor pressure:
33 mmHg

Product Name

Isopropanol, 70% in H2O

vapor density

2.1 (vs air)

Quality Level

vapor pressure

33 mmHg

form

liquid

expl. lim.

2-13.4 %

concentration

69-71 wt. % (2-propanol via KFB method)
70% in H2O

bp

80.9-83.2 °C

mp

-89.5 °C

density

0.858 g/mL at 25 °C

SMILES string

CC(C)O

InChI

1S/C3H8O/c1-3(2)4/h3-4H,1-2H3

InChI key

KFZMGEQAYNKOFK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Isopropanol, also known as 2-propanol or Isopropyl alcohol, is a 70% solution in water. i-PrOH is a secondary alcohol, colorless liquid, widely used as a solvent and intermediate in organic synthesis, miscible in solvents such as water, ethanol, ether, ethyl acetate, THF, and chloroform.

Application

Isopropanol can be used in many different applications due to its properties.
  • Solvent in organic synthesis
  • Chemical intermediate

Disclaimer

The product is not intended for use as a biocide under global biocide regulations, including but not limited to US EPA′s Federal Insecticide Fungicide and Rodenticide Act, European Biocidal Products Regulation, Canada’s Pest Management Regulatory Agency, Turkey’s Biocidal Products Regulation, Korea’s Consumer Chemical Products and Biocide Safety Management Act (K-BPR) and others.

greener alternative product

Product No.
Description
Pricing

related product

Product No.
Description
Pricing

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

72.0 °F - closed cup

Flash Point(C)

22.2 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Solubility of benzoic acid in acetone, 2-propanol, acetic acid and cyclohexane: Experimental measurement and thermodynamic modeling.
Long B, et al.
Fluid Phase Equilibria, 297(1), 113-120 (2010)
Influence of activated carbon in TiO2 and ZnO mediated photo-assisted degradation of 2-propanol in gas-solid regime.
Matos J, et al.
Applied Catalysis. B, Environmental, 99(1), 170-180 (2010)
Photocatalytic degradation of 2-propanol and phenol using Au loaded MnWO4 nanorod under visible light irradiation.
Chakraborty AK, et al.
Journal of Cluster Science, 23(2), 437-448 (2012)
Photocatalytic degradation of 2-propanol under irradiation of visible light by nanocrystalline titanium (IV) oxide modified with rhodium ion using adsorption method.
Kitano S, et al.
Chemistry Letters (Jpn), 39(6), 627-629 (2010)
2-propanol oxidation in a pressurized jet-stirred reactor (JSR) and combustion bomb: experimental and detailed kinetic modeling study.
Togbe C, et al.
Energy and Fuels, 25(2), 676-683 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service