Skip to Content
Merck
All Photos(3)

Documents

909955

Sigma-Aldrich

2-Propanol

greener alternative

≥99.5%, BioRenewable, ReagentPlus®

Synonym(s):

sec-Propyl alcohol, IPA, Isopropanol, Isopropyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHOH
CAS Number:
Molecular Weight:
60.10
Beilstein:
635639
MDL number:
UNSPSC Code:
12352104
NACRES:
NA.05

product name

2-Propanol, BioRenewable, ReagentPlus®, ≥99.5%

vapor density

2.1 (vs air)

vapor pressure

33 mmHg ( 20 °C)
44 mmHg ( 25 °C)

product line

ReagentPlus®

Assay

≥99.5%

form

liquid

autoignition temp.

750 °F

expl. lim.

2.0-12.7 %, 93 °C

greener alternative product characteristics

Safer Solvents and Auxiliaries
Use of Renewable Feedstocks
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

renewable carbon content

78%

refractive index

n20/D 1.377 (lit.)

bp

82 °C (lit.)

mp

−89.5 °C (lit.)

density

0.785 g/mL at 25 °C (lit.)

greener alternative category

SMILES string

CC(C)O

InChI

1S/C3H8O/c1-3(2)4/h3-4H,1-2H3

InChI key

KFZMGEQAYNKOFK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product is biorenewable and thus it aligns with "Safer Solvents and Auxiliaries" and "Use of Renewable Feedstocks". Click here  for more information. This ReagentPlus® grade 2-propanol is a drop-in replacement for any 2-propanol application. Made from 78.0% renewable carbon (verified through C14 ASTM D6866-16 testing) and produced in an environmentally friendly way that reduces the carbon footprint of production and reliance on fossil fuels. Because it is not from petroleum, it is free from many non-renewable contaminants, such as benzene, phthalates, and other phenols, leading to a higher quality product.

Application

Isopropanol can be used in different applications such as:
  • Solvent in organic synthesis
  • Chemical intermediate
  • Disinfectant
  • Antibacterial properties
  • Sample preservative

Features and Benefits

  • Renewable Carbon Content >78.0%
  • ASTM D6866 - Standard Test Methods for Determining the Biobased Content
  • Made from Renewable Resource - corn waste
  • Replacement for Petroleum-based 2-Propanol

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

53.6 °F - closed cup

Flash Point(C)

12.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service