Skip to Content
Merck
All Photos(3)

Key Documents

190764

Sigma-Aldrich

2-Propanol

≥99.5%, ACS reagent, suitable for RNA extraction

Synonym(s):

isopropanol, sec-Propyl alcohol, IPA, Isopropanol, Isopropyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHOH
CAS Number:
Molecular Weight:
60.10
Beilstein:
635639
EC Number:
MDL number:
UNSPSC Code:
12352104
PubChem Substance ID:
NACRES:
NA.21
grade:
ACS reagent
technique(s):
RNA extraction: suitable
bp:
82 °C (lit.)
vapor pressure:
33 mmHg ( 20 °C)
44 mmHg ( 25 °C)
Pricing and availability is not currently available.

Product Name

2-Propanol, ACS reagent, ≥99.5%

grade

ACS reagent

Quality Level

vapor density

2.1 (vs air)

vapor pressure

33 mmHg ( 20 °C)
44 mmHg ( 25 °C)

Assay

≥99.5%

form

liquid

autoignition temp.

750 °F

expl. lim.

2.0-12.7 %, 93 °C

technique(s)

RNA extraction: suitable

impurities

≤0.0001 meq/g Titr. acid or base
≤0.002% carbonyl compounds (as propionaldehyde or acetone)
≤0.2% water

evapn. residue

≤0.001%

color

APHA: ≤10

refractive index

n20/D 1.377 (lit.)

pH

7.0 (20 °C)

bp

82 °C (lit.)

mp

−89.5 °C (lit.)

solubility

H2O: passes test

density

0.785 g/mL at 25 °C (lit.)

SMILES string

CC(C)O

InChI

1S/C3H8O/c1-3(2)4/h3-4H,1-2H3

InChI key

KFZMGEQAYNKOFK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-propanol, also known as isopropanol or Isopropyl alcohol, is a secondary alcohol, colorless liquid, widely used as a solvent and intermediate in organic syntheses, miscible in solvents such as water, ethanol, ether, ethyl acetate, THF, and chloroform.

Application

Isopropanol can be used in many different applications due to its properties
  • Solvent in organic synthesis
  • Chemical intermediate

Features and Benefits

ACS solvents meet or exceed the high standards of the American Chemical Society (ACS) ,with test specifications that are specialized to every compound. According to the American Chemical Society, ACS reagent grade implies that it is a substance of sufficient purity to be used in most chemical analyses or reactions.

Disclaimer

The product is not intended for use as a biocide under global biocide regulations, including but not limited to US EPA′s Federal Insecticide Fungicide and Rodenticide Act, European Biocidal Products Regulation, Canada’s Pest Management Regulatory Agency, Turkey’s Biocidal Products Regulation, Korea’s Consumer Chemical Products and Biocide Safety Management Act (K-BPR) and others.

greener alternative product

Product No.
Description
Pricing

related product

Product No.
Description
Pricing

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

53.6 °F - closed cup

Flash Point(C)

12.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michelle T Paulsen et al.
Methods (San Diego, Calif.), 67(1), 45-54 (2013-08-27)
Gene expression studies commonly examine total cellular RNA, which only provides information about its steady-state pool of RNA. It remains unclear whether differences in the steady-state reflects variable rates of transcription or RNA degradation. To specifically monitor RNA synthesis and
Felix Moltzahn et al.
Journal of visualized experiments : JoVE, (54)(54), doi:10-doi:10 (2011-08-19)
The broad involvement of miRNAs in critical processes underlying development, tissue homoeostasis and disease has led to a surging interest among the research and pharmaceutical communities. To study miRNAs, it is essential that the quantification of microRNA levels is accurate
A study of anatase-supported Mn oxide as catalysts for 2-propanol oxidation.
Gallardo-Amores JM, et al.
Applied Catalysis. B, Environmental, 22(4), 249-259 (1999)
Highly efficient synthesis of cyclic ureas from CO2 and diamines by a pure CeO2 catalyst using a 2-propanol solvent.
Tamura M, et al.
Green Chemistry, 15(6), 1567-1577 (2013)
Electrochemical photocatalytic degradation of dye solution with a TiO2-coated stainless steel electrode prepared by electrophoretic deposition.
Lin WC, et al.
Applied Catalysis. B, Environmental, 140, 32-41 (2013)

Questions

1–4 of 4 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  3. Is 190764 - 2-Propanol sterile or non-sterile?

    1 answer
    1. This product has not been filtered and is not considered sterile.

      Helpful?

  4. The description for this chemical is somewhat confusing. It states it's a 70% solution in water even though the spec sheet for this product is >99% pure. What is exactly meant by this.

    1 answer
    1. Thank you for calling this to our attention. This product is a pure or absolute alcohol. It has not been diluted. A request to correct this statement has been submitted.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service