Skip to Content
Merck
All Photos(2)

Key Documents

34867

Sigma-Aldrich

1-Butanol

suitable for HPLC, ≥99.7%

Synonym(s):

n-Butanol, Butyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3OH
CAS Number:
Molecular Weight:
74.12
Beilstein:
969148
EC Number:
MDL number:
UNSPSC Code:
41116105
PubChem Substance ID:
NACRES:
NA.21
grade:
HPLC grade
technique(s):
HPLC: suitable
UV/Vis spectroscopy: suitable
application(s):
food and beverages
bp:
116-118 °C (lit.)
vapor pressure:
5.5 mmHg
Pricing and availability is not currently available.

grade

HPLC grade

Quality Level

vapor density

2.55 (vs air)

vapor pressure

5.5 mmHg

Assay

≥99.7%

form

liquid

autoignition temp.

649 °F

expl. lim.

11.2 %

technique(s)

HPLC: suitable
UV/Vis spectroscopy: suitable

impurities

≤0.001% non-volatile matter
≤0.002% free acid (as C3H7COOH)
≤0.1% water (Karl Fischer)

transmittance

210 nm, ≥20%
235 nm, ≥80%
280 nm, ≥98%

refractive index

n20/D 1.399 (lit.)

pH

7 (20 °C, 70 g/L)

bp

116-118 °C (lit.)

mp

−90 °C (lit.)

density

0.81 g/mL at 25 °C (lit.)

application(s)

food and beverages

SMILES string

CCCCO

InChI

1S/C4H10O/c1-2-3-4-5/h5H,2-4H2,1H3

InChI key

LRHPLDYGYMQRHN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-Butanol is a linear alcohol. Its biosynthesis from glucose using a bioengineered Escherichia coli strain has been reported.[1] Its reaction with diisocyanates in the presence of triethylamine (catalyst) has been investigated by volumetric method. Kinetic studies suggest that the reaction proceeds via competitive consecutive second order.[2] Its dehydration reaction has been employed as a standard reaction to evaluate the acidic and basic characteristics of various silica-aluminas.[3]

Application

1-Butanol may be used for the solid acid (an acidic salt of H3PW12O40, Cs2.5H0.5PW12O40) catalyzed esterification of acrylic acid.[4] It may be used as a cosolvent to investigate the enantioselective separation of racemic cyclohexyl methylphosphonofluoridate (cyclosarin, GF) by an chiral supercritical fluid chromatographic (SFC) technique.[5]
Suitable for HPLC, spectrophotometry, environmental testing

Packaging

M-Bottle for Solvents
As a global leader in lab reagents, we are constantly looking for new ways to optimize the safety of our products. The newly developed 4L solvent bottle design features advanced sealing technology that eliminates leaks to make the handling of solvents safer and more convenient than ever before.
See all the new features here!

Other Notes

Important notice
The article number 34867-4X2.5L will be discontinued. Please order the single bottle 34867-2.5L which is physically identical with the same exact specifications.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

95.0 °F - Pensky-Martens closed cup

Flash Point(C)

35 °C - Pensky-Martens closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The kinetics of the triethylamine-catalyzed reaction of diisocyanates with 1-butanol in toluene.
Burkus J and Eckert CF.
Journal of the American Chemical Society, 80(22), 5948-5950 (1958)
Acid-base properties of silica-aluminas: use of 1-butanol dehydration as a test reaction.
Berteau P, et al.
Applied Catalysis, 70(1), 307-323 (1991)
Shane A Kasten et al.
Chirality, 26(12), 817-824 (2014-10-10)
Chemical warfare nerve agents (CWNAs) are extremely toxic organophosphorus compounds that contain a chiral phosphorus center. Undirected synthesis of G-type CWNAs produces stereoisomers of tabun, sarin, soman, and cyclosarin (GA, GB, GD, and GF, respectively). Analytical-scale methods were developed using
Liquid phase esterification of acrylic acid with 1-butanol catalyzed by solid acid catalysts.
Chen X, et al.
Applied Catalysis A: General, 180(!), 261-269 (1999)
C R Shen et al.
Metabolic engineering, 10(6), 312-320 (2008-09-09)
Production of higher alcohols via the keto-acid intermediates found in microorganism's native amino-acid pathways has recently shown promising results. In this work, an Escherichia coli strain that produces 1-butanol and 1-propanol from glucose was constructed. The strain first converts glucose

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service