Skip to Content
Merck
All Photos(1)

Key Documents

1.01988

Supelco

1-Butanol

for liquid chromatography LiChrosolv®

Synonym(s):

1-Butanol, Butan-1-ol, n-Butyl alcohol, BuOH

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3OH
CAS Number:
Molecular Weight:
74.12
MDL number:
UNSPSC Code:
12191502
EC Index Number:
200-751-6
NACRES:
NA.03
Assay:
≥99.8% (GC)
technique(s):
HPLC: suitable
bp:
116-118 °C/1013 hPa
vapor pressure:
6.7 hPa ( 20 °C)
Pricing and availability is not currently available.

vapor pressure

6.7 hPa ( 20 °C)

Quality Level

product line

LiChrosolv®

grade

isocratic

Assay

≥99.8% (GC)

form

liquid

autoignition temp.

340 °C

potency

790 mg/kg LD50, oral (Rat)
3400 mg/kg LD50, skin (Rabbit)

expl. lim.

1.4-11.3 % (v/v)

technique(s)

HPLC: suitable

impurities

≤0.0002 meq/g Acidity
≤0.0002 meq/g Alkalinity
≤0.05% Water

evapn. residue

≤2.0 mg/L

transmittance

230 nm, ≥75%
240 nm, ≥85%
310 nm, ≥99%

pH

7 (20 °C, 70 g/L in H2O)

bp

116-118 °C/1013 hPa

mp

-89 °C

transition temp

flash point 34 °C

solubility

66 g/L

density

0.81 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

OCCCC

InChI

1S/C4H10O/c1-2-3-4-5/h5H,2-4H2,1H3

InChI key

LRHPLDYGYMQRHN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

With their high degree of UV transmittance, low particle count, low acidity and alkalinity and low evaporation residue level, LiChrosolv solvents are ideal for reproducible separations. Since separations are normally carried out under gradient conditions in analytical HPLC, we offer solvents in "gradient grade" as well as "isocratic grade". This enables to minimize the gradient effect of the solvent involved.

Application


  • Insights into "wheat aroma": Analysis of volatile components in wheat grains cultivated in saline-alkali soil: This study explores the use of 1-butanol in analyzing the aromatic profiles of wheat grains from saline-alkali soils, highlighting its role in identifying key volatile compounds associated with the unique scent of wheat (Sun et al., 2024).

  • PLD2 deficiency alleviates endothelial glycocalyx degradation in LPS-induced ARDS/ALI: Includes findings on the protective role of 1-butanol in cellular models of lung injury, demonstrating its potential therapeutic applications in respiratory conditions (Kong et al., 2024).


Preparation Note

Product filtered through a 0.2 μm filter

Other Notes

Explore various lab safety accessories and equipment for safe handling of solvents to increase your safety level from the first usage.

Legal Information

LICHROSOLV is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

95.0 °F - Pensky-Martens closed cup

Flash Point(C)

35 °C - Pensky-Martens closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service