Skip to Content
Merck
All Photos(3)

Key Documents

294810

Sigma-Aldrich

2-Butanol

anhydrous, 99.5%

Synonym(s):

sec-Butyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH(OH)CH3
CAS Number:
Molecular Weight:
74.12
Beilstein:
773649
EC Number:
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.21
grade:
anhydrous
Assay:
99.5%
bp:
98 °C (lit.)
vapor pressure:
12.5 mmHg ( 20 °C)

grade

anhydrous

Quality Level

vapor density

2.6 (vs air)

vapor pressure

12.5 mmHg ( 20 °C)

Assay

99.5%

form

liquid

autoignition temp.

761 °F

expl. lim.

9.8 %

impurities

<0.005% water

evapn. residue

<0.0003%

refractive index

n20/D 1.397 (lit.)

bp

98 °C (lit.)

mp

−115 °C (lit.)

density

0.808 g/mL at 25 °C (lit.)

SMILES string

CCC(C)O

InChI

1S/C4H10O/c1-3-4(2)5/h4-5H,3H2,1-2H3

InChI key

BTANRVKWQNVYAZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2-Butanol may be used in butyl lithium tritation to check its molarity using 1,10-phenanthroline as indicator.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

80.6 °F - closed cup

Flash Point(C)

27 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jiafeng Lu et al.
Oncology letters, 10(6), 3457-3465 (2016-01-21)
The aim of the present study was to obtain comprehensive microRNA (miRNA) profiles of type I [Ishikawa (ISK)] and type II (HEC-1B) human endometrial adenocarcinoma cell lines, utilizing the latest high-throughput sequencing techniques. RNA was extracted from ISK and HEC-1B
Selective Ruthenium-Catalyzed Transfer Hydrogenations of Nitriles to Amines with 2-Butanol.
Cai J, et al.
Combustion and Flame, 160(10), 1939-1957 (2013)
1-Phenyl-2,3,4,5-tetramethylphosphole
Fagan PJ and Nugent WA
Organic Syntheses, 70, 272-272 (1992)
Genesis of Br?nsted acid sites during dehydration of 2-butanol on tungsten oxide catalysts.
Baertsch CD, et al.
J. Catal., 205(1), 44-57 (2002)
The microcatalytic technique applied to a zero order reaction: the dehydration of 2-butanol over hydroxyapatite catalysts.
Bett JAS and Hall WK.
J. Catal., 10(2), 105-113 (1968)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service