Skip to Content
Merck
All Photos(1)

Key Documents

33117

Sigma-Aldrich

Cyclohexane

puriss. p.a., ACS reagent, ≥99.5% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12
CAS Number:
Molecular Weight:
84.16
Beilstein:
1900225
EC Number:
MDL number:
UNSPSC Code:
12191502
PubChem Substance ID:
NACRES:
NA.21
Assay:
≥99.5% (GC)
bp:
80.7 °C (lit.)
vapor pressure:
168.8 mmHg ( 37.7 °C)
77 mmHg ( 20 °C)
Pricing and availability is not currently available.

grade

ACS reagent
puriss. p.a.

Quality Level

vapor density

2.9 (vs air)

vapor pressure

168.8 mmHg ( 37.7 °C)
77 mmHg ( 20 °C)

Assay

≥99.5% (GC)

form

liquid

autoignition temp.

500 °F

expl. lim.

9 %

impurities

≤0.001% free acid (as HCl)
≤0.001% non-volatile matter
≤0.01% water (Karl Fischer)
≤0.05% aromatic substances (as C6H6)
≤0.05% cyclohexene (C6H10)

color

APHA: ≤10

refractive index

n20/D 1.4250-1.4280
n20/D 1.426 (lit.)

bp

80.7 °C (lit.)

mp

4-7 °C (lit.)

transition temp

solidification point 5.5-6.5 °C

density

0.779 g/mL at 25 °C (lit.)

cation traces

Al: ≤0.5 mg/kg
B: ≤0.02 mg/kg
Ba: ≤0.1 mg/kg
Ca: ≤0.5 mg/kg
Cd: ≤0.05 mg/kg
Co: ≤0.02 mg/kg
Cr: ≤0.02 mg/kg
Cu: ≤0.02 mg/kg
Fe: ≤0.1 mg/kg
Mg: ≤0.1 mg/kg
Mn: ≤0.02 mg/kg
Ni: ≤0.02 mg/kg
Pb: ≤0.1 mg/kg
Sn: ≤0.1 mg/kg
Zn: ≤0.1 mg/kg

suitability

passes test for reaction against H2SO4

SMILES string

C1CCCCC1

InChI

1S/C6H12/c1-2-4-6-5-3-1/h1-6H2

InChI key

XDTMQSROBMDMFD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Cyclohexane undergoes oxidation in the presence of various oxidants such as hydrogen peroxide, tert-butyl hydroperoxide and molecular oxygen.[1]

Application

Cyclohexane has been used to prepare a 1.3M solution of sec-butyllithium.[2] It may be used as a starting reagent for the synthesis of adipic acid[3] and ε-caprolactam.[3]

Other Notes

The article number 33117-4X2.5L will be discontinued. Please order the single bottle 33117-2.5L which is physically identical with the same exact specifications.
The article number 33117-4X2.5L-M will be discontinued. Please order the single bottle 33117-2.5L-M which is physically identical with the same exact specifications.
Pertains only in Germany: unversteuert

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

-4.0 °F - closed cup

Flash Point(C)

-20 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hyperbranched nanomolecules: Regular polystyrene dendrigrafts.
Muchtar Z, et al.
Macromolecules, 34(22), 7595-7600 (2001)
Cyclohexane oxidation continues to be a challenge.
Schuchardt ULF, et al.
Applied Catalysis A: General, 211(1), 1-17 (2001)
Eagleson M.
Concise Encyclopedia Chemistry, 168-168 (1994)
Amanda L Pitts et al.
Journal of the American Chemical Society, 136(24), 8614-8625 (2014-05-16)
Carbon-hydrogen bond activation reactions of four cycloalkanes (C5H10, C6H12, C7H14, and C8H16) by the Cp'Rh(CO) fragments (Cp' = η(5)-C5H5 (Cp) or η(5)-C5Me5 (Cp*)) were modeled theoretically by combining density functional and coupled cluster theories, and their reaction rates were measured
Robert Duran et al.
Environmental science and pollution research international, 22(20), 15215-15229 (2015-02-26)
The exploration of marine Actinobacteria has as major challenge to answer basic questions of microbial ecology that, in turn, will provide useful information to exploit Actinobacteria metabolisms in biotechnological processes. The ecological functions performed by Actinobacteria in marine sediments are

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service