Skip to Content
Merck
All Photos(3)

Documents

179191

Sigma-Aldrich

Cyclohexane

ACS reagent, ≥99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12
CAS Number:
Molecular Weight:
84.16
Beilstein:
1900225
EC Number:
MDL number:
UNSPSC Code:
12191502
eCl@ss:
39010606
PubChem Substance ID:
NACRES:
NA.21

grade

ACS reagent

Quality Level

vapor density

2.9 (vs air)

vapor pressure

168.8 mmHg ( 37.7 °C)
77 mmHg ( 20 °C)

Assay

≥99%

form

liquid

autoignition temp.

500 °F

expl. lim.

9 %

impurities

H2SO4, passes test (darkened)
≤0.02% water

evapn. residue

≤0.002%

color

APHA: ≤10

refractive index

n20/D 1.426 (lit.)

bp

80.7 °C (lit.)

mp

4-7 °C (lit.)

density

0.779 g/mL at 25 °C (lit.)

SMILES string

C1CCCCC1

InChI

1S/C6H12/c1-2-4-6-5-3-1/h1-6H2

InChI key

XDTMQSROBMDMFD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Cyclohexane is a cyclic alkane that predominantly exists in chair conformation due to higher stability. It participates as starting reagent in the photonitrosylation process (PNC process) and affords ε-caprolactum. Its oxidation by employing various oxidants such as hydrogen peroxide, tert-butyl hydroperoxide and molecular oxygen has been reported.

Application

Cyclohexane has been used in the following studies:
  • As a solvent in the synthesis of europium dibenzoylmethide triethylammonium (EuD4TEA).
  • Preparation of poly(dimethylsiloxane) (PDMS) microchip.
  • As a solvent in the two-solvent (cyclohexane/water) deposition method for the preparation of monometallic 5wt% Ni containing catalysts.
Cyclohexane may undergo oxidation using hydrogen peroxide as oxidant and in the presence of a polyoxotungstate catalyst to form cyclohexanone and cyclohexanol as the main products. Its dehydrogenation in the presence of Ni-Cu/SiO2 catalysts forms benzene in good yield.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

-4.0 °F - closed cup

Flash Point(C)

-20 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Samuel Kim et al.
Methods in enzymology, 472, 119-132 (2010-06-29)
Microfluidics serves as a convenient platform for single-molecule experiments by providing manipulation of small amounts of liquids and micron-sized particles. An adapted version of capillary electrophoresis (CE) on a microchip can be utilized to separate chemical species with high resolution
Characterizations and performances of Ni/diatomite catalysts for dry reforming of methane.
Jabbour K, et al.
Chemical Engineering Journal, 264, 351-358 (2015)
Cyclohexane dehydrogenation over Ni-Cu/SiO 2 catalyst: Effect of copper addition.
Xia Z, et al.
Catalysis Communications, 90, 39-42 (2017)
The energy difference between the chair and boat forms of cyclohexane. The twist conformation of Cyclohexane.
Johnson WS, et al.
Journal of the American Chemical Society, 83(3), 606-614 (1961)
Cyclohexane oxidation continues to be a challenge.
Schuchardt ULF, et al.
Applied Catalysis A: General, 211(1), 1382-1386 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service