Skip to Content
Merck
All Photos(1)

Documents

32249

Sigma-Aldrich

Toluene

puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.7% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH3
CAS Number:
Molecular Weight:
92.14
Beilstein:
635760
EC Number:
MDL number:
UNSPSC Code:
12352005
eCl@ss:
39011102
PubChem Substance ID:
NACRES:
NA.21

grade

ACS reagent
puriss. p.a.

Quality Level

Agency

USP/NF
reag. ISO
reag. Ph. Eur.

vapor density

3.2 (vs air)

vapor pressure

22 mmHg ( 20 °C)
26 mmHg ( 25 °C)

Assay

≥99.7% (GC)

form

liquid

autoignition temp.

997 °F

expl. lim.

7 %

technique(s)

microbiological culture: suitable

impurities

≤0.0001% thiophene
≤0.001% free acid (as HCl)
≤0.001% free alkali (as NaOH)
≤0.001% non-volatile matter
≤0.003% S-compounds (as S)
≤0.02% water (Karl Fischer)

color

APHA: ≤10

refractive index

n/D 1.496 (lit.)
n20/D 1.496-1.498

bp

110-111 °C (lit.)

mp

-93 °C (lit.)

density

0.865 g/mL at 25 °C (lit.)

cation traces

Al: ≤0.5 ppm
Ba: ≤0.1 ppm
Bi: ≤0.1 ppm
Ca: ≤0.5 ppm
Cd: ≤0.05 ppm
Co: ≤0.02 ppm
Cr: ≤0.02 ppm
Cu: ≤0.02 ppm
Fe: ≤0.1 ppm
K: ≤0.5 ppm
Li: ≤0.1 ppm
Mg: ≤0.1 ppm
Mn: ≤0.02 ppm
Mo: ≤0.1 ppm
Na: ≤0.5 ppm
Ni: ≤0.02 ppm
Pb: ≤0.1 ppm
Sr: ≤0.1 ppm
Zn: ≤0.1 ppm

SMILES string

Cc1ccccc1

InChI

1S/C7H8/c1-7-5-3-2-4-6-7/h2-6H,1H3

InChI key

YXFVVABEGXRONW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Toluene, a benzene analog, is widely employed as an organic solvent. Its anaerobic biodegradation to CO2, by the denitrifying bacterium Thauera arornatica has been reported. It forms syndiotactic polystyrene-toluene molecular compound. Crystal structure of this molecular compound has been investigated by X-ray diffraction studies.

Application

Toulene may be used as a solvent for the preparation of 4-arm polylactide-based (PLA) star oligomer. It was used for the activation of glass slides, for in vitro test system for allergy diagnosis.

Other Notes

The article number 32249-4X2.5L will be discontinued. Please order the single bottle 32249-2.5L which is physically identical with the same exact specifications.
The article number 32249-4X2.5L-M will be discontinued. Please order the single bottle 32249-2.5L-M which is physically identical with the same exact specifications.
The article number 32249-6X1L will be discontinued. Please order the single bottle 32249-1L which is physically identical with the same exact specifications.
The article number 32249-6X1L-M will be discontinued. Please order the single bottle 32249-1L-M which is physically identical with the same exact specifications.
Pertains only in Germany: unversteuert

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 Inhalation - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

39.9 °F - closed cup

Flash Point(C)

4.4 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Barbara I Fall et al.
Analytical chemistry, 75(3), 556-562 (2003-02-15)
The described in vitro test system for allergy diagnosis is based on microscope glass slides activated with (3-glycidyloxypropyl)trimethoxysilane. Allergen solutions are immobilized as small droplets (approximately 10 nL) on the activated glass slides with a piezoelectric arrayer. In contrast to
T Biegert et al.
European journal of biochemistry, 238(3), 661-668 (1996-06-15)
Toluene is degraded anoxically to CO2 by the denitrifying bacterium Thauera aromatica. Toluene first becomes oxidized to benzoyl-CoA by O2-independent reactions. Benzoyl-CoA is then reduced to non-aromatic products by benzoyl-CoA reductase. We set out to study the reactions employed for
Structural study on syndiotactic polystyrene: 2. Crystal structure of molecular compound with toluene.
Chatani Y, et al.
Polymer, 34(8), 1620-1620 (1993)
V Melissinaki et al.
Biofabrication, 3(4), 045005-045005 (2011-09-21)
This study reports on the production of high-resolution 3D structures of polylactide-based materials via multi-photon polymerization and explores their use as neural tissue engineering scaffolds. To achieve this, a liquid polylactide resin was synthesized in house and rendered photocurable via
Liang Guo et al.
Journal of chromatography. A, 1286, 9-15 (2013-03-19)
For the first time, a novel one-step sample preparation method that combines microwave assisted extraction and solvent bar microextraction (MAE-SBME) with analysis by gas chromatography-mass spectrometry (GC-MS), was developed for the fast and efficient determination of polycyclic aromatic hydrocarbons (PAHs)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service