Skip to Content
Merck
All Photos(8)

Key Documents

746355

Sigma-Aldrich

Zinc chloride

anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥97%

Synonym(s):

Dichlorozinc

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ZnCl2
CAS Number:
Molecular Weight:
136.30
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21
Assay:
≥97%
grade:
ACS reagent
anhydrous
form:
powder, crystals or granules

grade

ACS reagent
anhydrous

Quality Level

vapor pressure

1 mmHg ( 428 °C)

product line

Redi-Dri

Assay

≥97%

form

powder, crystals or granules

quality

free-flowing

reaction suitability

reagent type: catalyst
core: zinc

impurities

Oxychloride, passes test
≤0.005% insolubles

pH

5 (20 °C, 100 g/L)

mp

293 °C (lit.)

anion traces

nitrate (NO3-): ≤0.003%
sulfate (SO42-): ≤0.01%

cation traces

Ca: ≤0.06%
Fe: ≤0.001%
K: ≤0.02%
Mg: ≤0.01%
NH4+: ≤0.005%
Na: ≤0.05%
Pb: ≤0.005%

SMILES string

[Cl-].[Cl-].[Zn++]

InChI

1S/2ClH.Zn/h2*1H;/q;;+2/p-2

InChI key

JIAARYAFYJHUJI-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Zinc chloride (ZnCl2) is an inorganic compound that is commonly used as a reagent in chemical synthesis. It is used as a mild Lewis acid catalyst in cycloaddition, substitution, electrophilic aromatic additions, and selective reduction reactions. ZnCl2 is also a useful starting material for the synthesis of many organozinc reagents via transmetalation reaction.

Application

Zinc chloride can be used as:     
  • A catalyst to synthesize 5-substituted 1H-tetrazoles by [3 + 2] cycloaddition reaction of organic nitriles with sodium azide.      
  • A reagent to promote the hydrolysis of NaBH4 for the generation of hydrogen.      
  • A reagent for the activation of carbon materials.

Legal Information

Redi-Dri is a trademark of Sigma-Aldrich Co. LLC

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Guiran Xiao et al.
Cell reports, 26(3), 748-758 (2019-01-17)
Iron metabolism is an essential process that when dysregulated causes disease. Mammalian serum transferrin (TF) plays a primary role in delivering iron to cells. To improve our understanding of the conservation of iron metabolism between species, we investigate here the
E Hjortsø et al.
Intensive care medicine, 14(1), 17-24 (1988-01-01)
Five soldiers were injured by inhalation of hexite smoke (ZnCl2) during military training. Two soldiers, not wearing gas masks breathed hexite for 1 or 2 min, they slowly developed severe adult respiratory distress syndrome (ARDS) over the ensuing 2 weeks.
Use of unlicensed black salve for cutaneous malignancy.
Natalie C Ong et al.
The Medical journal of Australia, 200(6), 314-314 (2014-04-08)
Dadong Guo et al.
Archives of biochemistry and biophysics, 560, 44-51 (2014-07-23)
Zinc (Zn), the second abundant trace element in living organisms, plays an important role in regulating cell metabolism, signaling, proliferation, gene expression and apoptosis. Meanwhile, the overload of Zn will disrupt the intracellular calcium homeostasis via impairing mitochondrial function. However
Nathalie Adam et al.
Nanotoxicology, 8(7), 709-717 (2013-07-11)
In this study, the effect of ZnO nanoparticles and ZnCl2 on growth, reproduction and accumulation of zinc in Daphnia magna was determined in a 21-day chronic toxicity test. A variety of techniques were used to distinguish the free zinc ion

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service