Skip to Content
Merck
All Photos(1)

Key Documents

Y0001498

Lopinavir

European Pharmacopoeia (EP) Reference Standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C37H48N4O5
CAS Number:
Molecular Weight:
628.80
UNSPSC Code:
41116107
NACRES:
NA.24

biological source

synthetic

grade

pharmaceutical primary standard

Agency

EP

API family

lopinavir

packaging

pkg of 170 mg

manufacturer/tradename

EDQM

storage condition

protect from light

application(s)

pharmaceutical (small molecule)

format

neat

shipped in

ambient

storage temp.

2-8°C

SMILES string

N4(CCCNC4=O)[C@@H](C(C)C)C(=O)N[C@H](C[C@H](O)[C@@H](NC(=O)COc3c(cccc3C)C)Cc2ccccc2)Cc1ccccc1

InChI

1S/C37H48N4O5/c1-25(2)34(41-20-12-19-38-37(41)45)36(44)39-30(21-28-15-7-5-8-16-28)23-32(42)31(22-29-17-9-6-10-18-29)40-33(43)24-46-35-26(3)13-11-14-27(35)4/h5-11,13-18,25,30-32,34,42H,12,19-24H2,1-4H3,(H,38,45)(H,39,44)(H,40,43)/t30-,31-,32-,34-/m0/s1

InChI key

KJHKTHWMRKYKJE-SUGCFTRWSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.

Lopinavir is an antiretroviral agent and a protease inhibitor indicated for the management of human immunodeficiency virus (HIV) infection. It is a key component of Kaletra, the co-formulation of lopinavir and ritonavir.

Application

Lopinavir may be used as a EP reference standard for the determination of the analyte in pharmaceutical formulations and plasma samples by chromatography techniques.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of 19 antiretroviral agents in pharmaceuticals or suspected products with two methods using high-performance liquid chromatography
Rebiere H, et al.
Journal of Chromatography. B, Biomedical Applications, 850(1-2), 376-383 (2007)
Simultaneous determination of indinavir, ritonavir and lopinavir (ABT 378) in human plasma by high-performance liquid chromatography
Ray J, et al.
Journal of Chromatography. B, Biomedical Applications, 775(2), 225-230 (2002)
Lopinavir
European Pharmacopoeia Commission and European Directorate for the Quality of Medicines & Healthcare
European pharmacopoeia, 3(F), 3111-3111 (2020)
Therapeutic drug monitoring of lopinavir/ritonavir in pregnancy
Lambert JS, et al.
HIV Medicine, 12(3), 166-173 (2011)
Validation and application of a high-performance liquid chromatography-tandem mass spectrometric method for simultaneous quantification of lopinavir and ritonavir in human plasma using semi-automated 96-well liquid-liquid extraction
Wang PG, et al.
Journal of Chromatography A, 1130(2), 302-307 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service