Skip to Content
Merck
All Photos(1)

Key Documents

1270480

USP

Fingolimod Hydrochloride

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

FTY720, 2-Amino-2-[2-(4-octyl-phenyl)-ethyl]-propane-1,3-diol hydrochloride, Fingolimod hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H33NO2 · HCl
CAS Number:
Molecular Weight:
343.93
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

biological source

synthetic

Agency

USP

API family

fingolimod

form

powder

manufacturer/tradename

USP

storage condition

protect from light

color

white

solubility

DMSO: soluble
ethyl acetate: soluble
water: soluble

application(s)

pharmaceutical

storage temp.

2-8°C

SMILES string

Cl.NC(CO)(CO)CCc1ccc(cc1)CCCCCCCC

InChI

1S/C19H33NO2.ClH/c1-2-3-4-5-6-7-8-17-9-11-18(12-10-17)13-14-19(20,15-21)16-22;/h9-12,21-22H,2-8,13-16,20H2,1H3;1H

InChI key

SWZTYAVBMYWFGS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Fingolimod Hydrochloride (fingolimod HCl), an S1P-receptor modulator, belongs to the class of synthetic glucocorticoid steroid drugs.
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets, have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Fingolimod Hydrochloride USP reference standard may be used to quantify the analyte in gel, capsule and other pharmaceutical formulations, active pharmaceutical ingredient (API) and pharmaceutical dosage forms by liquid chromatography (LC) based techniques.

Biochem/physiol Actions

FTY720 is an immunomodulating drug and sphingosine 1-phosphate (S1P) receptor modulator. Phosphorylation of FTY270 by sphingosine kinase causes S1P1R internalization, which sequesters lymphocytes in lymph nodes, preventing them from taking part in an autoimmune response. Clinically, it has been approved for the treatment of multiple sclerosis (MS). It has also been shown to block and reverse paclitaxel-induced chemotherapy induced peripheral neuropathy (CIPN) through S1PR inhibition as well as inhibit the activity of histone deacetylases in the hippocampus of mouse brains, thereby modulating memory.
FTY720, an immunomodulating drug, acts as a sphingosine 1-phosphate (S1P) receptor modulator.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

STOT RE 2

Target Organs

Immune system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fingolimod Hydrochloride
Fingolimod Hydrochloride USP Monograph; USP42-NF37:United States Pharmacopeia and National Formulary
United States Pharmacopeia, 43(6)(1), 1849-1849 (2020)
A randomized, open-label study assessing the bioequivalence of two formulations of Fingolimod 0.5 mg in healthy subjects
Tanguay M, et al.
AAPS Open, 4(1), 3-3 (2018)
Rapid determination of fingolimod hydrochloride-related Substances and degradation products in API and pharmaceutical dosage forms by use of a stability-indicating UPLC method
Rajan N and Basha KA
Chromatographia, 77(21-22), 1545-1552 (2014)
Solubility of dexamethasone in supercritical carbon dioxide
Chim RB, et al.
Journal of Chemical and Engineering Data, 57(12), 3756-3760 (2012)
Sphingosine 1-phosphate receptor 1 as a useful target for treatment of multiple sclerosis
Chiba K and Adachi K
Pharmaceuticals (Basel, Switzerland), 5(5), 514-528 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service