Skip to Content
Merck
All Photos(5)

Key Documents

T4375

Sigma-Aldrich

Tetrazolium Blue Chloride

used in colorimetric determination of reducing compounds

Synonym(s):

3,3′-(3,3′-Dimethoxy[1,1′-biphenyl]-4,4′-diyl)-bis(2,5-diphenyl-2H-tetrazolium) dichloride, 3,3′-Dianisole-4,4′-bis(3,5-diphenyltetrazolium chloride), BTC, Blue Tetrazolium chloride, TZ

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C40H32Cl2N8O2
CAS Number:
Molecular Weight:
727.64
Beilstein:
3894077
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

Quality Level

form

powder

mp

255 °C (dec.) (lit.)

solubility

methanol: 50 mg/mL, clear

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[Cl-].[Cl-].COc1cc(ccc1-[n+]2nc(nn2-c3ccccc3)-c4ccccc4)-c5ccc(c(OC)c5)-[n+]6nc(nn6-c7ccccc7)-c8ccccc8

InChI

1S/C40H32N8O2.2ClH/c1-49-37-27-31(23-25-35(37)47-43-39(29-15-7-3-8-16-29)41-45(47)33-19-11-5-12-20-33)32-24-26-36(38(28-32)50-2)48-44-40(30-17-9-4-10-18-30)42-46(48)34-21-13-6-14-22-34;;/h3-28H,1-2H3;2*1H/q+2;;/p-2

InChI key

MUUHXGOJWVMBDY-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Tetrazolium blue chloride (BTC) is a colorless tetrazolium compound that upon reduction forms a colored diformazan. The reactivity has been applied to the analysis of reducing compounds including the nanomolar detection of reducing sugars and carbidopa and methyldopa. BTC has been used for histological staining of mouse coronal brain sections and rat brain sections. In combination with 4-nitrocatechol, tetrazolium blue chloride forms a ternary complex with molybdenum(VI) for extraction and spectrophotometric determination of molybdenum. It has also been used to stain Francisella tularensis cells for tularemic microagglutination test.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Preparation of a new tetrazolium salt which yields a blue pigment on reduction and its use in the demonstration of enzymes in normal and neoplastic tissues.
A M RUTENBURG et al.
Cancer research, 10(2), 113-121 (1950-02-01)
C K Jue et al.
Journal of biochemical and biophysical methods, 11(2-3), 109-115 (1985-08-01)
Addition of sodium potassium tartrate to basic solutions of tetrazolium blue [(2,2',5,5'-tetraphenyl-3,3'-dimethoxy 4,4'-biphenylene) ditetrazolium chloride] greatly improved their efficacy as colorimetric reagents for reducing sugar determination. This modification increased sensitivity and decreased reaction time. The modified reagent can determine as
Sensitive colorimetric assay of carbidopa and methyldopa using tetrazolium blue chloride in pharmaceutical products.
Issopoulos, P.B.
International Journal of Clinical Pharmacy, 11, 213-217 (1989)
[Development of a novel Francisella tularensis antigen stained with tetrazolium-blue for tularemia microagglutination test].
Celebi B
Mikrobiyoloji bulteni (2013)
Ternary complex of molybdenum(VI) with 4-nitrocatechol and tetrazolium blue chloride and Its application to extraction-spectrophotometric analysis of ferrous metallurgy products.
Dimitrov., A.N., et al.
Journal of Analytical Chemistry, 62, 122-125 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service