Skip to Content
Merck
All Photos(1)

Documents

11585029001

Roche

NBT

crystals, pkg of 5 g

Synonym(s):

Nitrotetrazolium Blue chloride, 2,2′-bis(4-Nitrophenyl)-5,5′-diphenyl-3,3′-(3,3′-dimethoxy-4,4′-diphenylene)ditetrazolium chloride, 3,3′-(3,3′-Dimethoxy-4,4′-biphenylene)bis[2-(4-nitrophenyl)-5-phenyl-2H-tetrazolium chloride], p-Nitro-Blue tetrazolium chloride, p-Nitrotetrazolium blue, NBT, Nitro BT

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C40H30N10O6 · 2Cl
CAS Number:
Molecular Weight:
817.64
Beilstein:
4115923
MDL number:
UNSPSC Code:
41116100
PubChem Substance ID:

description

4-Nitro blue tetrazolium chloride, crystals

Quality Level

form

crystals

packaging

pkg of 5 g

manufacturer/tradename

Roche

storage temp.

2-8°C

SMILES string

[Cl-].[Cl-].COc1cc(ccc1-[n+]2nc(nn2-c3ccc(cc3)[N+]([O-])=O)-c4ccccc4)-c5ccc(c(OC)c5)-[n+]6nc(nn6-c7ccc(cc7)[N+]([O-])=O)-c8ccccc8

InChI

1S/C40H30N10O6.2ClH/c1-55-37-25-29(13-23-35(37)47-43-39(27-9-5-3-6-10-27)41-45(47)31-15-19-33(20-16-31)49(51)52)30-14-24-36(38(26-30)56-2)48-44-40(28-11-7-4-8-12-28)42-46(48)32-17-21-34(22-18-32)50(53)54;;/h3-26H,1-2H3;2*1H/q+2;;/p-2

InChI key

FSVCQIDHPKZJSO-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

NBT is soluble in water, aqueous buffers, and dimethylformamide.
Tetrazolium salts are useful in determining the metabolic activity of cells from many origins. The quaternary tetrazole ring core containing four nitrogen atoms, with a net positive charge induces cellular uptake through plasma membrane potential. This drives its use in cell biology. In immunolabeling technique, 4-Nitro blue tetrazolium (NBT) serves as an oxidant. The hydrolysis of 5-bromo-4-chloro-3-indolyl phosphate (BCIP) by alkaline phosphatase gives rise to leucoindigo, which is then being reduced by NBT to form an insoluble blue BCI. Certain cells reduce NBT, which is a colorless/yellow dye to blue or black formazan crystals. Due to this property, NBT is prefered in redox histochemistry and in biochemical applications. It is formerly used to distinguish between nonbacterial and bacterial diseases, the latter causing neutrophils to reduce the dye; used to confirm diagnosis of chronic granulomatous disease.

Application

  • Colony and plaque hybridization
  • Immunohistocytochemistry
  • In situ hybridization
  • Northern blot
  • Southern blot
  • Western blot
NBT has been used in in situ hybridization.

Quality

Performance-controlled in the LDH assay.

Preparation Note

Working solution: Solubility: soluble in water, aqueous buffers, and dimethylformamide. Can be dissolved in methanol. A clear solution is achieved at 20 mg/ml.

Analysis Note

Absorption: NBT has its maximum absorption at 260 nm [in water],
and the final product formazan has its maximum absorption at 605 nm [in nitrobenzole].

Other Notes

For life science research only. Not for use in diagnostic procedures.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tetrazolium dyes as tools in cell biology: new insights into their cellular reduction.
Berridge M V, et al.
Biotechnology Annual Review, 11, 127-152 (2005)
Infection and nitroblue-tetrazolium reduction by neutrophils: A diagnostic aid.
Park B H, et al.
Lancet, 292(7567), 532-534 (1968)
D Liebhart et al.
Journal of comparative pathology, 135(4), 237-242 (2006-10-31)
For use in an in-situ hybridization method, three probes (HM, TR and BL) were designed to hybridize, respectively, with (1) Histomonas meleagridis, (2) Tetratrichomonas gallinarum, and (3) a broad range of micro-organisms, including Blastocystis spp. Mono-eukaryotic cultures were used to
Fluorescent in situ hybridization employing the conventional NBT/BCIP chromogenic stain.
Trinh L A, et al.
Biotechniques, 42(6), 756-759 (2007)
Wanlong Su et al.
Frontiers in plant science, 11, 568411-568411 (2021-02-09)
Salt stress is an adverse environmental factor for plant growth and development. Under salt stress, plants can activate the selective autophagy pathway to alleviate stress. However, the regulatory mechanism of selective autophagy in response to salt stress remains largely unclear.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service