Skip to Content
Merck
All Photos(8)

Documents

N6876

Sigma-Aldrich

Nitrotetrazolium Blue chloride

≥90.0% (HPLC)

Synonym(s):

2,2′-bis(4-Nitrophenyl)-5,5′-diphenyl-3,3′-(3,3′-dimethoxy-4,4′-diphenylene)ditetrazolium chloride, 3,3′-(3,3′-Dimethoxy-4,4′-biphenylene)bis[2-(4-nitrophenyl)-5-phenyl-2H-tetrazolium chloride], p-Nitro-Blue tetrazolium chloride, p-Nitrotetrazolium blue, NBT, Nitro BT

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C40H30N10O6 · 2Cl
CAS Number:
Molecular Weight:
817.64
Beilstein:
4115923
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

Quality Level

Assay

≥90.0% (HPLC)

form

powder

impurities

≤10% solvent

solubility

H2O: 10 mg/mL

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

2-8°C

SMILES string

[Cl-].[Cl-].COc1cc(ccc1-[n+]2nc(nn2-c3ccc(cc3)[N+]([O-])=O)-c4ccccc4)-c5ccc(c(OC)c5)-[n+]6nc(nn6-c7ccc(cc7)[N+]([O-])=O)-c8ccccc8

InChI

1S/C40H30N10O6.2ClH/c1-55-37-25-29(13-23-35(37)47-43-39(27-9-5-3-6-10-27)41-45(47)31-15-19-33(20-16-31)49(51)52)30-14-24-36(38(26-30)56-2)48-44-40(28-11-7-4-8-12-28)42-46(48)32-17-21-34(22-18-32)50(53)54;;/h3-26H,1-2H3;2*1H/q+2;;/p-2

InChI key

FSVCQIDHPKZJSO-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

  • Nitrotetrazolium Blue chloride has been used to detect the generation of oxygen metabolites in cells.
  • It has been used as a substrate for western blot detection.
  • It has been used for the staining of histological tissue sections.

Biochem/physiol Actions

Nitrotetrazolium blue (NBT), in combination with BCIP (5-bromo-4-chloro-3-indolyl-phosphate), is required for the detection of labeled probes in western blots. It is part of the colorimetric detection assay. NBT and BCIP dyes detect alkaline phosphatase activity, thereby generating an insoluble purple dye (diformazan). The dye is visible at areas where the labeled probe has hybridized to its target.

Substrates

Substrate for dehydrogenases and other oxidases.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 2 - STOT SE 3

Target Organs

Eyes,Central nervous system, Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fluorescent in situ hybridization employing the conventional NBT/BCIP chromogenic stain.
Trinh le A et al.
Biotechniques, 42, 756-756 (2007)
Methods in Enzymology, 6, 958-958 (1963)
Expression of the endothelial lipase gene in murine embryos and reproductive organs.
Lindegaard ML et al.
Journal of Lipid Research, 46, 439-439 (2005)
Drought- and salt-tolerant plants result from overexpression of the AVP1 H+-pump.
Gaxiola RA et al.
Proceedings of the National Academy of Sciences of the USA, 98, 11444-11444 (2001)
Measurement of reactive oxygen intermediate production in haemocytes of the penaeid shrimp, Penaeus ?annamei.
Knaap V et al.
Aquaculture (Amsterdam, Netherlands), 191 , 89-107 (2000)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service