Skip to Content
Merck
All Photos(1)

Key Documents

N2125

Sigma-Aldrich

Naphthol AS-BI phosphate

Naphthol AS-BI phosphate

≥98% purity (HPLC), powder

Synonym(s):

7-Bromo-3-hydroxy-2-naphthoic-o-anisidide phosphate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H15BrNO6P
CAS Number:
Molecular Weight:
452.19
Beilstein:
6668526
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47
Pricing and availability is not currently available.

Product Name

Naphthol AS-BI phosphate, ≥98% (HPLC), Technical grade

Quality Level

Assay

≥98% (HPLC)

form

powder

technique(s)

microbe id | staining: suitable

color

white to light yellow

solubility

methanol: 50 mg/mL

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

−20°C

SMILES string

COc1ccccc1NC(=O)c2cc3cc(Br)ccc3cc2OP(O)(O)=O

InChI

1S/C18H15BrNO6P/c1-25-16-5-3-2-4-15(16)20-18(21)14-9-12-8-13(19)7-6-11(12)10-17(14)26-27(22,23)24/h2-10H,1H3,(H,20,21)(H2,22,23,24)

InChI key

HUXIAXQSTATULQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Naphthol AS-BI, also known as 7-Bromo-3-hydroxy-2-naphtho-o-anisidine is a substituted naphthol.

Application

Naphthol AS-BI phosphate is suitable to use:
  • in tartrate-resistant acid phosphatase (TRAP) staining to detect osteoclast activity
  • as a component in Tris-HCl in staining cells to measure alkaline phosphatase (ALP) activity
  • to perform alkaline phosphatase reaction
Substrate for the histochemical demonstration of acid and alkaline phosphatase. Substrate for the fluorometric assay of acid and alkaline phosphatases. Demonstration of acid phosphatase in paraffin embedded tissue. Cytofluorometric assay of acid phosphatase in single living cells.

Biochem/physiol Actions

Naphthol AS-BI can be used as a substrate to determine alkaline and acid phosphatase. It is also used to determine the activity of acid phosphatase in single cells.

Substrates

Substrate for the histochemical demonstration of acid and alkaline phosphatase.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Quantitative histochemical investigations of semipermeable membrane techniques for the assay of acid phosphatase in skeletal muscle. I. Meijer's method.
P J Stoward et al.
Histochemistry, 71(3), 405-431 (1981-01-01)
M Kimura et al.
Histochemistry, 82(6), 519-523 (1985-01-01)
A cytochemical study of acid phosphatase (AcP-ase) in the lateral prostate of the rat was performed to investigate whether AcP-ase in the secretory apparatus can be distinguished from AcP-ase in lysosomes and their related structures. Two types of AcP-ase were
Anthony J Janckila et al.
Clinica chimica acta; international journal of clinical chemistry, 347(1-2), 157-167 (2004-08-18)
Our purpose was to develop a specific immunoassay for tartrate-resistant acid phosphatase (TRACP) 5b using naphthol ASBI phosphate (N-ASBI-P) as a selective substrate for isoform 5b and heparin as a selective inhibitor of isoform 5a. Serum TRACP 5a and 5b
A M Boesen
Scandinavian journal of haematology, 32(3), 245-252 (1984-03-01)
The subcellular localization of acid phosphatase (AcP) in various immunologically-defined neoplastic lymphoid cells including hairy cells was investigated by electron microscopy. 2 substrates, naphthol-AS-BI-phosphoric acid (naphthol-AS-BI-P) and sodium beta-glycerophosphate, were compared. By incubation in naphthol-AS-BI-P containing medium, the reaction product
A J Janckila et al.
Journal of bone and mineral research : the official journal of the American Society for Bone and Mineral Research, 16(4), 788-793 (2001-04-24)
Tartrate-resistant acid phosphatase (TRAP) isoform 5b is a potential serum marker for osteoclastic activity. Biochemical assays for serum TRAP activity with para-nitrophenylphosphate (pNPP) have low specificity for bone because of hydrolysis by unrelated nontype 5 TRAPs of blood cells and

Questions

Reviews

Active Filters

  1. 1 Ratings-Only Review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service