Skip to Content
Merck
All Photos(2)

Documents

SCR004

Sigma-Aldrich

Alkaline Phosphatase Detection Kit

This Alkaline Phosphatase Detection Kit is a specific & sensitive tool for the phenotypic assessment of Embryonic Stem (ES) cell differentiation by the determination of AP activity.

Synonym(s):

ALP Stain, Alkaline Phosphtase Stem Cell Staining Kit, Stem Cell AP Staining Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
eCl@ss:
32161000
NACRES:
NA.84

Quality Level

species reactivity (predicted by homology)

mammals

manufacturer/tradename

Chemicon®

technique(s)

cell culture | stem cell: suitable

input

sample type cardiac stem cell(s)
sample type: mouse embryonic stem cell(s)
sample type hematopoietic stem cell(s)
sample type induced pluripotent stem cell(s)
sample type neural stem cell(s)
sample type epithelial cells
sample type pancreatic stem cell(s)
sample type mesenchymal stem cell(s)
sample type: human embryonic stem cell(s)

shipped in

wet ice

General description

Millipore′s Alkaline Phosphatase Detection Kit (Catalog number SCR004) is a specific and sensitive tool for the phenotypic assessment of ES cell differentiation by the determination of AP activity.

Application

Embryonic stem (ES) cells are totipotent cells derived from the inner cell mass (ICM) of preimplantation mammalian embryos and are capable of unlimited, undifferentiated proliferation in vitro [1,2,3]. Undifferentiated murine ES cells can be maintained in vitro for extended periods in media containing the cytokine, leukemia-inhibitory factor (LIF) or Millipore′s proprietary ES cell culture reagent, ESGRO [4,5]. The undifferentiated state of ES cells can be characterized by high level of expression of Alkaline Phosphatase (AP) [6], the expression of surface markers including SSEA and TRA antigens and the transcription factor Oct-4.

Millipore′s Alkaline Phosphatase Detection Kit (Catalog number SCR004) is a specific and sensitive tool for the phenotypic assessment of ES cell differentiation by the determination of AP activity.

Available separately from Millipore are the monoclonal antibodies TRA-2-49 (Catalog Number MAB4349) and TRA-2-54 (Catalog Number MAB4354), which permit the detection of Liver / Bone / Kidney isozyme of AP as expressed by ES cells. When used in flow cytometry, these reagents permit a quantitative assessment of AP expression by ES cells [7].

For Research Use Only; Not for use in diagnostic procedures
Research Category
Stem Cell Research

Components

Fast Red Violet solution (0.8g/L stock) (Part No. 90239). Two 15mL bottles.

Napthol AS-BI phosphate solution (4mg/mL) in AMPD buffer (2mol/L), pH 9.5 (Part No. 90234). One 15mL bottle.

Storage and Stability

The Alkaline Phosphatase Detection Kit consists of two components used for determining AP activity.

When stored at 2-8º C, the kit components are stable up to the expiration date. Do not freeze or expose to elevated temperatures. Discard any remaining reagents after the expiration date.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ahmed K Elsayed et al.
Stem cell research, 42, 101705-101705 (2020-01-29)
FOXA2 is a transcription factor, playing an important role during development. We established an induced pluripotent stem cell (iPSC) line, QBRIi009-A, using non-integrating Sendai virus from a 4-year-old boy, displaying a complex clinical phenotype. Molecular karyotyping and cytogenetics confirmed a
Jinkyung Cho et al.
The Journal of nutritional biochemistry, 77, 108327-108327 (2020-01-12)
Self-renewal is required for embryo stem cells (ESCs) and adipose-derived mesenchymal stem cells (ADMSCs). This study examined the ability of ferulic acid in mouse ESCs and ADMSCs, in a high fat diet-induced mouse model. Initially, five natural compounds of ferulic
Efficient iPS cell production with the MyoD transactivation domain in serum-free culture.
Hiroyuki Hirai,Nobuko Katoku-Kikyo,Peter Karian,Meri Firpo,Nobuaki Kikyo
Testing null
Feeder-independent culture of mouse embryonic stem cells using vitamin A/retinol.
Jaspal S Khillan,Liguo Chen
Methods in Molecular Biology null
Efficient feeder-free episomal reprogramming with small molecules.
Junying Yu,Kevin Fongching Chau,Maxim A Vodyanik,Jinlan Jiang,Yong Jiang
Testing null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service