Skip to Content
Merck
All Photos(3)

Documents

A5144

Sigma-Aldrich

8-Anilino-1-naphthalenesulfonic acid hemimagnesium salt hydrate

≥95% purity (perchloric acid titration), for fluorescence, powder

Synonym(s):

1,8-ANS magnesium salt, ANSA magnesium salt, Magnesium 8-anilino-1-naphthalenesulfonate hydrate, N-Phenyl peri acid magnesium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H12Mg0.5NO3S · xH2O
CAS Number:
Molecular Weight:
310.49 (anhydrous basis)
Beilstein:
4834623
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

product name

8-Anilino-1-naphthalenesulfonic acid hemimagnesium salt hydrate, for fluorescence, ≥95% (TLC)

grade

for fluorescence

Quality Level

Assay

≥95% (TLC)

form

powder

color

faint green to green

solubility

H2O: 50 mg/mL

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

O.[Mg++].[O-]S(=O)(=O)c1cccc2cccc(Nc3ccccc3)c12.[O-]S(=O)(=O)c4cccc5cccc(Nc6ccccc6)c45

InChI

1S/2C16H13NO3S.Mg.H2O/c2*18-21(19,20)15-11-5-7-12-6-4-10-14(16(12)15)17-13-8-2-1-3-9-13;;/h2*1-11,17H,(H,18,19,20);;1H2/q;;+2;/p-2

InChI key

FBANPIUXAXTAFX-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

ANSA (8-Anilino-1-naphthalenesulfonic acid) is a hydrophobic polarity sensitive fluorescent dye useful as a site probe to detect conformational changes in cell and micelle membranes and molecules such as proteins. ANSA is also used as a fluorescent probe for the estimation of the critical micelle concentration (CMC) of surfactants.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rebecca L Smathers et al.
PloS one, 7(6), e38459-e38459 (2012-06-16)
4-Hydroxynonenal (4-HNE) is a reactive α,β-unsaturated aldehyde produced during oxidative stress and subsequent lipid peroxidation of polyunsaturated fatty acids. The reactivity of 4-HNE towards DNA and nucleophilic amino acids has been well established. In this report, using proteomic approaches, liver
Stefan Kofler et al.
Journal of molecular biology, 422(1), 109-123 (2012-05-29)
The ability of pathogenesis-related proteins of family 10 to bind a broad spectrum of ligands is considered to play a key role for their physiological and pathological functions. In particular, Bet v 1, an archetypical allergen from birch pollen, is
Chang Li et al.
International journal of biological macromolecules, 49(5), 910-916 (2011-08-23)
In our study, we showed that at a relatively low concentration, H(2)O(2) can irreversibly inactivate the human brain type of creatine kinase (HBCK) and that HBCK is inactivated in an H(2)O(2) concentration-dependent manner. HBCK is completely inactivated when incubated with
Shilpi Gupta et al.
Small (Weinheim an der Bergstrasse, Germany), 9(6), 894-904 (2012-12-12)
A new class of non-ionic dendronized multiamphiphilic polymers is prepared from a biodegradable (AB)n-type diblock polymer synthesized from 2-azido-1,3-propanediol (azido glycerol) and polyethylene glycol (PEG)-600 diethylester using Novozym-435 (Candida antarctica lipase) as a biocatalyst, following a well-established biocatalytic route. These
Silvia Zorrilla et al.
Methods in molecular biology (Clifton, N.J.), 952, 237-252 (2012-10-27)
Transcription factors of the peroxisome proliferator-activated receptor (PPAR) family are ligand-activated receptors that play key roles in lipid metabolism and inflammation. The γ isoform (PPARγ) is involved in adipocyte differentiation, insulin sensitization, and vascular pathophysiology, including inflammation and atherosclerosis, for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service