Skip to Content
Merck
All Photos(1)

Documents

10416

Sigma-Aldrich

8-Anilino-1-naphthalenesulfonic acid ammonium salt

technical, ≥90% (NT)

Synonym(s):

1,8-ANS NH4, ANSA, Ammonium 8-anilino-1-naphthalenesulfonate, N-Phenyl peri acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C16H13NO3S · NH3
CAS Number:
Molecular Weight:
316.37
Beilstein:
3581235
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.32

grade

technical

Quality Level

Assay

≥90% (NT)

form

powder

color

, Green to Dark Green and Grey to Dark Grey and Green-Grey

solubility

NaOH: 1 N
H2O: soluble
methanol: soluble

fluorescence

λex 388 nm; λem 470 nm in 0.1 M Tris, 0.2 M KCl, pH 9.0, BSA

SMILES string

N.OS(=O)(=O)c1cccc2cccc(Nc3ccccc3)c12

InChI

1S/C16H13NO3S.H3N/c18-21(19,20)15-11-5-7-12-6-4-10-14(16(12)15)17-13-8-2-1-3-9-13;/h1-11,17H,(H,18,19,20);1H3

InChI key

IPBNQYLKHUNLQE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

ANS forms an inclusion complex with cyclodextrin. Such model systems are useful to mimic biological recognition and can be studied by measuring the change in fluorescence of free-ANS to complexed-ANS. When ANS enters the hydrophobic core of cyclodestrin, it′s fluorescence increases . Utilized in the reagent phase of a sodium-selective fiber-optic sensor. The reagent phase also contains a copper(II) polyelectrolyte, which binds to ANSA in the absence of sodium and quenches the fluorescence. In the presence of sodium, ANSA forms a cationic complex creating ion-pairs, causing it to fluoresce . ANS is often incorporated into di-block polymers and can be released by changes in the local environment (i.e., temperature, pH, etc.) . ANS is commonly used as a fluorescence probe to investigate molecular assemblies of surfactants and amphiphilic polymers because a blue shift of the emission maximum indicates the fluorophore is located in less polar media . Fluorescent probe for protein studies using methodologies such as steady-state and dynamic fluorescence measurements .

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rebecca L Smathers et al.
PloS one, 7(6), e38459-e38459 (2012-06-16)
4-Hydroxynonenal (4-HNE) is a reactive α,β-unsaturated aldehyde produced during oxidative stress and subsequent lipid peroxidation of polyunsaturated fatty acids. The reactivity of 4-HNE towards DNA and nucleophilic amino acids has been well established. In this report, using proteomic approaches, liver
Beiwen Zheng et al.
PloS one, 6(5), e19510-e19510 (2011-05-26)
The scavenging ability of sufficient divalent metal ions is pivotal for pathogenic bacteria to survive in the host. ATP-binding cassette (ABC)-type metal transporters provide a considerable amount of different transition metals for bacterial growth. TroA is a substrate binding protein
Chang Li et al.
International journal of biological macromolecules, 49(5), 910-916 (2011-08-23)
In our study, we showed that at a relatively low concentration, H(2)O(2) can irreversibly inactivate the human brain type of creatine kinase (HBCK) and that HBCK is inactivated in an H(2)O(2) concentration-dependent manner. HBCK is completely inactivated when incubated with
Dimitra Georgiadou et al.
PloS one, 6(11), e27037-e27037 (2011-11-10)
Apolipoprotein E (apoE) is a major protein of the lipoprotein transport system that plays important roles in lipid homeostasis and protection from atherosclerosis. ApoE is characterized by structural plasticity and thermodynamic instability and can undergo significant structural rearrangements as part
Shilpi Gupta et al.
Small (Weinheim an der Bergstrasse, Germany), 9(6), 894-904 (2012-12-12)
A new class of non-ionic dendronized multiamphiphilic polymers is prepared from a biodegradable (AB)n-type diblock polymer synthesized from 2-azido-1,3-propanediol (azido glycerol) and polyethylene glycol (PEG)-600 diethylester using Novozym-435 (Candida antarctica lipase) as a biocatalyst, following a well-established biocatalytic route. These

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service