Merck
  • Hydrogen peroxide targets the cysteine at the active site and irreversibly inactivates creatine kinase.
Hydrogen peroxide targets the cysteine at the active site and irreversibly inactivates creatine kinase.
International journal of biological macromolecules (2011-08-23)
Chang Li, Shiqing Sun, Daeui Park, Hyoung Oh Jeong, Hae Young Chung, Xiao-Xia Liu, Hai-Meng Zhou
ABSTRACT
In our study, we showed that at a relatively low concentration, H(2)O(2) can irreversibly inactivate the human brain type of creatine kinase (HBCK) and that HBCK is inactivated in an H(2)O(2) concentration-dependent manner. HBCK is completely inactivated when incubated with 2mM H(2)O(2) for 1h (pH 8.0, 25°C). Inactivation of HBCK is a two-stage process with a fast stage (k(1)=0.050 ± 0.002 min(-1)) and a slow (k(2)=0.022 ± 0.003 min(-1)) stage. HBCK inactivation by H(2)O(2) was affected by pH and therefore we determined the pH profile of HBCK inactivation by H(2)O(2). H(2)O(2)-induced inactivation could not be recovered by reducing agents such as dl-dithiothreitol, N-acetyl-L-cysteine, and l-glutathione reduced. When HBCK was treated with DTNB, an enzyme substrate that reacts specifically with active site cysteines, the enzyme became resistant to H(2)O(2). HBCK binding to Mg(2+)ATP and creatine can also prevent H(2)O(2) inactivation. Intrinsic and 1-anilinonaphthalene-8-sulfonate-binding fluorescence data showed no tertiary structure changes after H(2)O(2) treatment. The thiol group content of H(2)O(2)-treated HBCK was reduced by 13% (approximately 1 thiol group per HBCK dimer, theoretically). For further insight, we performed a simulation of HBCK and H(2)O(2) docking that suggested the CYS283 residue could interact with H(2)O(2). Considering these results and the asymmetrical structure of HBCK, we propose that H(2)O(2) specifically targets the active site cysteine of HBCK to inactivate HBCK, but that substrate-bound HBCK is resistant to H(2)O(2). Our findings suggest the existence of a previously unknown negative form of regulation of HBCK via reactive oxygen species.
Sigma-Aldrich
8-Anilino-1-naphthalenesulfonic acid
Sigma-Aldrich
8-Anilino-1-naphthalenesulfonic acid ammonium salt, technical, ≥90% (NT)
Sigma-Aldrich
8-Anilino-1-naphthalenesulfonic acid ammonium salt, for fluorescence, ≥97.0% (HPLC)
Sigma-Aldrich
8-Anilino-1-naphthalenesulfonic acid hemimagnesium salt hydrate, for fluorescence, ≥95.0% (T)
Sigma-Aldrich
8-Anilino-1-naphthalenesulfonic acid hemimagnesium salt hydrate, for fluorescence, ≥95% (TLC)