Skip to Content
Merck
All Photos(3)

Documents

A0843

Sigma-Aldrich

Aluminum sulfate

meets USP testing specifications

Synonym(s):

Aluminum sulfate hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Al2(SO4)3 · xH2O
CAS Number:
Molecular Weight:
342.15 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21

Agency

USP/NF
meets USP testing specifications

Quality Level

form

powder

density

1.69 g/mL at 25 °C (lit.)

application(s)

pharmaceutical (small molecule)

SMILES string

[Al+3].[Al+3].[H]O[H].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

InChI

1S/2Al.3H2O4S.H2O/c;;3*1-5(2,3)4;/h;;3*(H2,1,2,3,4);1H2/q2*+3;;;;/p-6

InChI key

BUACSMWVFUNQET-UHFFFAOYSA-H

Looking for similar products? Visit Product Comparison Guide

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Najla A Lakkis et al.
Wilderness & environmental medicine, 26(3), 422-429 (2015-05-04)
Jellyfish have a worldwide distribution. Their stings can cause different reactions, ranging from cutaneous, localized, and self-limited to serious systemic or fatal ones, depending on the envenoming species. Several first aid treatments are used to manage such stings but few
Sandra G Velleman et al.
Avian diseases, 59(3), 410-418 (2015-10-20)
The wooden breast condition is a myopathy affecting the pectoralis major (p. major) muscle in fast-growing commercial broiler lines. Currently, wooden breast-affected birds are phenotypically detected by palpation of the breast area, with affected birds having a very hard p.
S Jolly et al.
Environmental science and pollution research international, 21(22), 13103-13117 (2014-07-06)
Aluminium is used in diverse anthropogenic processes at the origin of pollution events in aquatic ecosystems. In the Champagne region (France), high concentrations of aluminium (Al) are detected due to vine-growing practices. In fish, little is known about the possible
K Stölzel et al.
Cell and tissue banking, 16(1), 159-170 (2014-05-17)
Adult mesenchymal stem cells (MSCs) are a promising cell source in tissue engineering due to their availability, ease of isolation and high proliferative activity. This study was undertaken to investigate whether immortalised human MSC are able to undergo chondrogenic differentiation
Alonso G P Guedes et al.
American journal of physiology. Lung cellular and molecular physiology, 308(5), L485-L493 (2015-01-13)
CD38 is a cell-surface protein involved in calcium signaling and contractility of airway smooth muscle. It has a role in normal airway responsiveness and in airway hyperresponsiveness (AHR) developed following airway exposure to IL-13 and TNF-α but appears not to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service