Skip to Content
Merck
All Photos(4)

Documents

M7506

Sigma-Aldrich

Magnesium sulfate

anhydrous, ReagentPlus®, ≥99.5%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
MgSO4
CAS Number:
Molecular Weight:
120.37
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21

grade

anhydrous

Agency

suitable for SM 5210

vapor density

<0.01 (vs air)

vapor pressure

<0.1 mmHg ( 20 °C)

product line

ReagentPlus®

Assay

≥99.5%

form

powder

pH

~7.9 (25 °C, 50 g/L)

SMILES string

[Mg++].[O-]S([O-])(=O)=O

InChI

1S/Mg.H2O4S/c;1-5(2,3)4/h;(H2,1,2,3,4)/q+2;/p-2

InChI key

CSNNHWWHGAXBCP-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Magnesium sulfate is a magnesium salt with tocolytic ability. Its role to induce an analgesic effect during intraoperative and postoperative periods has been investigated. It is reported to prevent eclampsia. Its ability to remove water from water-miscible solvents by salting-out phase separation has been studied.

Application

Magnesium sulfate (anhydrous) can be used as a catalyst:      
  • In Biginelli reactions to synthesize dihydropyrimidinones by solvent-free conditions.      
  • In the synthesis of 2-imidazolines by the reaction of aldehydes and ethylenediamine in the presence of hydrogen peroxide and sodium iodide.

The mixture of MgSO4 and bayerite is used to prepare single-phase,MgAl2O4 spinel nanoparticles by calcining the powder mixture at 800 oC.
MgSO4 (anhydrous) can also be used as a drying agent in organic synthesis for the removal of traces of water from organic extracts.
Magnesium sulfate may be used as a drying agent for organic extracts.

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany
Redi-Dri is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A facile and efficient synthesis of 2-imidazolines from aldehydes using hydrogen peroxide and substoichiometric sodium iodide
Bai Guo-yi, et al.
Synthesis, 2011(10), 1599-1603 (2011)
Anhydrous magnesium sulfate mediated solvent-free synthesis of dihydropyrimidin-2 (1H)-ones at ambient temperature
Pore DM, et al.
Australian Journal of Chemistry, 60(6), 435-438 (2007)
Synthesis of MgAl2O4 spinel nanoparticles using a mixture of bayerite and magnesium sulfate
Su Xinghua, et al.
Journal of Nanoparticle Research, 12(5), 1813-1819 (2010)
A comparison of magnesium sulfate and nimodipine for the prevention of eclampsia.
Belfort MA, et al.
The New England Journal of Medicine, 348(4), 304-311 (2003)
H Koinig et al.
Anesthesia and analgesia, 87(1), 206-210 (1998-07-14)
In a randomized, double-blind study with two parallel groups, we assessed the analgesic effect of perioperative magnesium sulfate administration in 46 ASA physical status I or II patients undergoing arthroscopic knee surgery with total i.v. anesthesia. The patients received either

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service