Merck
  • Airway responsiveness in CD38-deficient mice in allergic airway disease: studies with bone marrow chimeras.
Airway responsiveness in CD38-deficient mice in allergic airway disease: studies with bone marrow chimeras.
American journal of physiology. Lung cellular and molecular physiology (2015-01-13)
Alonso G P Guedes, Joseph A Jude, Jaime Paulin, Laura Rivero-Nava, Hirohito Kita, Frances E Lund, Mathur S Kannan
ABSTRACT
CD38 is a cell-surface protein involved in calcium signaling and contractility of airway smooth muscle. It has a role in normal airway responsiveness and in airway hyperresponsiveness (AHR) developed following airway exposure to IL-13 and TNF-α but appears not to be critical to airway inflammation in response to the cytokines. CD38 is also involved in T cell-mediated immune response to protein antigens. In this study, we assessed the contribution of CD38 to AHR and inflammation to two distinct allergens, ovalbumin and the epidemiologically relevant environmental fungus Alternaria. We also generated bone marrow chimeras to assess whether Cd38(+/+) inflammatory cells would restore AHR in the CD38-deficient (Cd38(-/-)) hosts following ovalbumin challenge. Results show that wild-type (WT) mice develop greater AHR to inhaled methacholine than Cd38(-/-) mice following challenge with either allergen, with comparable airway inflammation. Reciprocal bone marrow transfers did not change the native airway phenotypic differences between WT and Cd38(-/-) mice, indicating that the lower airway reactivity of Cd38(-/-) mice stems from Cd38(-/-) lung parenchymal cells. Following bone marrow transfer from either source and ovalbumin challenge, the phenotype of Cd38(-/-) hosts was partially reversed, whereas the airway phenotype of the WT hosts was preserved. Airway inflammation was similar in Cd38(-/-) and WT chimeras. These results indicate that loss of CD38 on hematopoietic cells is not sufficient to prevent AHR and that the magnitude of airway inflammation is not the predominant underlying determinant of AHR in mice.
Sigma-Aldrich
Pancuronium bromide
Pancuronium bromide, European Pharmacopoeia (EP) Reference Standard
Pancuronium bromide for system suitability, European Pharmacopoeia (EP) Reference Standard
Isoflurane, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Aluminum sulfate, 99.99% trace metals basis
Sigma-Aldrich
Aluminum sulfate, meets USP testing specifications