Skip to Content
Merck
All Photos(2)

Documents

95283

Supelco

Water

for HPCE, for luminescence, suitable for UV/Vis spectroscopy

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2O
CAS Number:
Molecular Weight:
18.02
Beilstein:
2050024
EC Number:
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:
NACRES:
NB.21

grade

for HPCE
for luminescence

Quality Level

technique(s)

UV/Vis spectroscopy: suitable

refractive index

n20/D 1.34 (lit.)

bp

100 °C (lit.)

mp

0 °C (lit.)

density

1.000 g/mL at 3.98 °C (lit.)

absorption

in accordance at 200-800 nm

absorbance ratio

A200 nm ≤0.05 (absorption measured against Suprasil-Standard)

suitability

in accordance for fluorescence emission

SMILES string

O

InChI

1S/H2O/h1H2

InChI key

XLYOFNOQVPJJNP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Water is widely used for HPCE (High-Performance Capillary Electrophoresis), luminescence and UV-spectroscopic studies.

Application

Water has been used for the conditioning of capillaries during the micellar electrokinetic capillary chromatographic analysis.

Preparation Note

prepared under clean room conditions and filtered through a 0.2 μm membrane filter

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Matteo Bonoli et al.
Journal of agricultural and food chemistry, 51(5), 1141-1147 (2003-02-20)
Antioxidant properties and stimulating effects of green tea are related to its content of cathechins and xanthines; tea quality evaluation is based on organoleptic tests and on the presence of those components. In this work, by a MEKC method, eight
Mary A Rodgers et al.
The Journal of experimental medicine, 211(7), 1333-1347 (2014-06-25)
Linear ubiquitination is a newly discovered posttranslational modification that is currently restricted to a small number of known protein substrates. The linear ubiquitination assembly complex (LUBAC), consisting of HOIL-1L, HOIP, and Sharpin, has been reported to activate NF-κB-mediated transcription in
Susu Duan et al.
Nature communications, 5, 5029-5029 (2014-10-10)
Oseltamivir-resistant H1N1 influenza viruses carrying the H275Y neuraminidase mutation predominated worldwide during the 2007-2009 seasons. Although several neuraminidase substitutions were found to be necessary to counteract the adverse effects of H275Y, the order and impact of evolutionary events involved remain
Hervé Marie-Nelly et al.
Nature communications, 5, 5695-5695 (2014-12-18)
Closing gaps in draft genome assemblies can be costly and time-consuming, and published genomes are therefore often left 'unfinished.' Here we show that genome-wide chromosome conformation capture (3C) data can be used to overcome these limitations, and present a computational
M Lieveld et al.
Virchows Archiv : an international journal of pathology, 465(6), 703-713 (2014-10-12)
Giant cell tumor of bone (GCTB) displays worrisome clinical features such as local recurrence and occasionally metastatic disease which are unpredictable by morphology. Additional routinely usable biomarkers do not exist. Gene expression profiles of six clinically defined groups of GCTB

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service