Merck
  • Gene expression profiling of giant cell tumor of bone reveals downregulation of extracellular matrix components decorin and lumican associated with lung metastasis.
Gene expression profiling of giant cell tumor of bone reveals downregulation of extracellular matrix components decorin and lumican associated with lung metastasis.
Virchows Archiv : an international journal of pathology (2014-10-12)
M Lieveld, E Bodson, G De Boeck, B Nouman, A M Cleton-Jansen, E Korsching, M S Benassi, P Picci, G Sys, B Poffyn, N A Athanasou, P C W Hogendoorn, R G Forsyth
ABSTRACT
Giant cell tumor of bone (GCTB) displays worrisome clinical features such as local recurrence and occasionally metastatic disease which are unpredictable by morphology. Additional routinely usable biomarkers do not exist. Gene expression profiles of six clinically defined groups of GCTB and one group of aneurysmal bone cyst (ABC) were determined by microarray (n = 33). The most promising differentially expressed genes were validated by Q-PCR as potential biomarkers in a larger patient group (n = 41). Corresponding protein expression was confirmed by immunohistochemistry. Unsupervised hierarchical clustering reveals a metastatic GCTB cluster, a heterogeneous, non-metastatic GCTB cluster, and a primary ABC cluster. Balanced score testing indicates that lumican (LUM) and decorin (DCN) are the most promising biomarkers as they have lower level of expression in the metastatic group. Expression of dermatopontin (DPT) was significantly lower in recurrent tumors. Validation of the results was performed by paired and unpaired t test in primary GCTB and corresponding metastases, which proved that the differential expression of LUM and DCN is tumor specific rather than location specific. Our findings show that several genes related to extracellular matrix integrity (LUM, DCN, and DPT) are differentially expressed and may serve as biomarkers for metastatic and recurrent GCTB.
Product Description
Pure Water Density Standard, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
Pure Water Density Standard, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
Sigma-Aldrich
Water, ACS reagent
Sigma-Aldrich
Water, HPLC Plus
Supelco
Water, for TOC analysis
Sigma-Aldrich
Water, BioPerformance Certified
Supelco
Water, ACS reagent, for ultratrace analysis
Supelco
Water, suitable for ion chromatography
Supelco
Water, for HPCE, for luminescence, suitable for UV/Vis spectroscopy
Sigma-Aldrich
Water, for cell biology, sterile ultrafiltered
Sigma-Aldrich
Water, deuterium-depleted, ≤1 ppm (Deuterium oxide)
Sigma-Aldrich
Water, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Water, PCR Reagent
Sigma-Aldrich
Water, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Water, Nuclease-Free Water, for Molecular Biology
Sigma-Aldrich
E-Toxate Water, endotoxin, free
Sigma-Aldrich
Water, suitable for HPLC
Supelco
Density Standard 998 kg/m3, H&D Fitzgerald Ltd. Quality
Sigma-Aldrich
Water, tested according to Ph. Eur.
Sigma-Aldrich
Water, Deionized
Sigma-Aldrich
Water, for molecular biology, sterile filtered
Sigma-Aldrich
Water-16O, ≥99.94 atom % 16O