Skip to Content
Merck
All Photos(2)

Key Documents

94035

Supelco

Chrysene

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H12
CAS Number:
Molecular Weight:
228.29
Beilstein:
1909297
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

448 °C (lit.)

mp

252-254 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

SMILES string

c1ccc2c(c1)ccc3c4ccccc4ccc23

InChI

1S/C18H12/c1-3-7-15-13(5-1)9-11-18-16-8-4-2-6-14(16)10-12-17(15)18/h1-12H

InChI key

WDECIBYCCFPHNR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Muta. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Theodoros Tzamkiozis et al.
Inhalation toxicology, 22 Suppl 2, 59-69 (2010-10-30)
This study presents different research techniques linked together to improve our understanding of the particulate matter (PM) impacts on health. PM samples from the exhaust of different vehicles were collected by a versatile aerosol concentration enrichment system (VACES). Waterborne PM
Bimal K Banik et al.
European journal of medicinal chemistry, 45(10), 4687-4691 (2010-08-13)
We describe herein the synthesis of novel 6,12-distributed chrysene as potent anticancer agents. In vitro and in vivo studies are also reported here.
José M Lorenzo et al.
Meat science, 89(1), 105-109 (2011-04-20)
The polycyclic aromatic hydrocarbon (PAH) content in two traditional smoked sausages from Spain was determined. Results showed, that total average levels of PAHs found were higher in "Chorizo de cebolla" (101.81 vs. 98.48 μgkg(-1)). Most contents of PAHs found in
Chirag M Ghevariya et al.
Bioresource technology, 102(20), 9668-9674 (2011-08-23)
Degradation of chrysene, a four ring High Molecular Weight (HMW) Polycyclic Aromatic Hydrocarbon (PAH) is of intense environmental interest, being carcinogenic, teratogenic and mutagenic. Multiple PAH degrading halotolerant Achromobacter xylosoxidans was isolated from crude oil polluted saline site. Response Surface
L Viñas et al.
Chemosphere, 75(4), 534-541 (2009-01-20)
Aliphatic and aromatic hydrocarbons were determined in three species of commercial shellfish, namely razor shells (Ensis arcuatus and Ensis siliqua), goose barnacle (Pollicipes cornucopia) and sea urchin (Paracentrotus lividus), living in different habitats and exhibiting different feeding behaviors. The samples

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service