Skip to Content
Merck
All Photos(2)

Key Documents

51968

Supelco

Benzo[a]pyrene

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

3,4-Benzopyrene, 3,4-Benzpyrene, Benzo[def]chrysene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H12
CAS Number:
Molecular Weight:
252.31
Beilstein:
1911333
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

storage condition

under inert gas (argon)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

495 °C (lit.)

mp

177-180 °C (lit.)

application(s)

environmental

format

neat

SMILES string

c1ccc2c(c1)cc3ccc4cccc5ccc2c3c45

InChI

1S/C20H12/c1-2-7-17-15(4-1)12-16-9-8-13-5-3-6-14-10-11-18(17)20(16)19(13)14/h1-12H

InChI key

FMMWHPNWAFZXNH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Benzo[a]pyrene is a high molecular weight, polycyclic aromatic hydrocarbon, which is formed the pyrolysis of organic matter, and are present in the environment particularly in the atmosphere as particulate matter.
This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

Ion Mobility: TWCCSN2 value of 147.9 Å2 [M+H]+

The collision cross section (CCS) measurement was provided by Waters Corporation, using the SYNAPT XS mass spectrometer.
For a description and overview of how ion mobility enables the measurement of the CCS of an ion visit ims.waters.com.
Further information on the SYNAPT XS mass spectrometer can be found on the IMS microsite and product webpage.

TWCCS measurements are expected to be within 2% of this reference value.

P/N 51968 is part of the Waters Extractables & Leachables UNIFI scientific library which can be downloaded from Waters Marketplace.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Muta. 1B - Repr. 1B - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

National air toxics program the integrated urban strategy, report to Congress (2000)
Bioaccumulation in Marine Organisms: Effect of Contaminants from Oil Well Produced Water (2002)
Yuji Fujita et al.
Bioorganic & medicinal chemistry, 18(3), 1194-1203 (2010-01-12)
The physiological role of aryl hydrocarbon receptor (AhR) is not yet fully understood, and investigation is hampered by the limited solubility of reported AhR ligands in aqueous media. To achieve improved solubility, we focused on our previous finding that planarity-disruption
M Auslander et al.
Marine biotechnology (New York, N.Y.), 10(3), 250-261 (2008-01-24)
Individuals of the fish Lithognathus mormyrus were exposed to a series of pollutants including: benzo[a]pyrene, pp-DDE, Aroclor 1254, perfluorooctanoic acid, tributyl-tin chloride, lindane, estradiol, 4-nonylphenol, methyl mercury chloride, and cadmium chloride. Five mixtures of the pollutants were injected. Each mixture
Gunnar Boysen et al.
Mutation research, 543(1), 17-30 (2003-01-03)
We review studies which investigate the presence, using structure-specific analytical methods, of DNA or protein adducts of the carcinogen benzo[a]pyrene (BaP) in human tissues. The analytical methods include high performance liquid chromatography with fluorescence detection and gas chromatography-mass spectrometry. Although

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service