Skip to Content
Merck
All Photos(1)

Key Documents

48563

Supelco

Benz[a]anthracene

analytical standard

Synonym(s):

1,2-Benzanthracene, Tetraphene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H12
CAS Number:
Molecular Weight:
228.29
Beilstein:
1909298
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

grade

analytical standard

CofA

current certificate can be downloaded

packaging

ampule of 100 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

437.6 °C (lit.)

mp

157-159 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

storage temp.

2-30°C

SMILES string

c1ccc2cc3c(ccc4ccccc34)cc2c1

InChI

1S/C18H12/c1-2-7-15-12-18-16(11-14(15)6-1)10-9-13-5-3-4-8-17(13)18/h1-12H

InChI key

DXBHBZVCASKNBY-UHFFFAOYSA-N

Gene Information

human ... CYP1A2(1544)

Looking for similar products? Visit Product Comparison Guide

General description

Benz[a]anthracene is a colorless plate like material, which belongs to the group of polycyclic aromatic hydrocarbon.

Application

Benz[a]anthracene may be used as an analytical standard for the determination of the analyte in human urine, marine samples and smoked fish by chromatography techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Estimation of Rate Constants for Gas-Phase Reactions of Chrysene, Benz [a] anthracene, and Benzanthrone with OH and NO3 Radicals via a Relative Rate Method in CCl4 Liquid Phase-System
Kameda T, et al.
Polycyclic Aromat. Compd., 37, 101-105 (2017)
Pohanish.PR
Sittig's Handbook of Toxic and Hazardous Chemicals and Carcinogens, (7) (2017)
The determination of polycyclic aromatic hydrocarbons in smoked fish by gas chromatography mass spectrometry with positive-ion chemical ionization
Yurchenko S and Molder U
Journal of food composition and analysis, 18(8), 857-869 (2005)
Determination of hydroxylated metabolites of polycyclic aromatic hydrocarbons in urine
Jongeneelen FJ, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 413, 227- 232 (1987)
Determination of polynuclear aromatic hydrocarbons in marine samples of Siokolo Fishing Settlement
Anyakora C, et al.
Journal of Chromatography A, 1073(1-2), 323-330 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service