Skip to Content
Merck
All Photos(1)

Key Documents

87972

Millipore

Peptone from soybean, enzymatic digest

for biotechnological purposes, Growth stimulant for fastidious microorganisms

Synonym(s):

Peptone from soybean

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
41106212
eCl@ss:
42040102
NACRES:
NA.85

biological source

soybean

Quality Level

grade

for biotechnological purposes

form

powder

composition

total nitrogen (N), ≥8%

storage condition

protect from light

ign. residue

≤22%

loss

≤6.0% loss on drying

pH

6.4-7.8

solubility

H2O: 2%, clear, brownish-yellow

application(s)

food and beverages
microbiology

storage temp.

2-30°C

Looking for similar products? Visit Product Comparison Guide

General description

Peptone from soybean is a papaic soy hydrolysate, multi-step purified with low molecular weight. It is widely used in the lactic acid bacteria fermentation industry. It is a fine beige powder easily soluble in water. It is manufactured with raw materials from vegetal origin only (non-GMO).It′s natural nitrogen content combined with naturally occurring vitamins as well as a high concentration of carbohydrates makes this peptone suitable for cultivating fastidious microorganisms.

Application

Peptone from soybean is used in the preparation of microbiological culture media. Owing to its rich nutrient source, it is ideal for facilitating growth of a variety of microorganisms including fastidious bacteria or fungi.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service