Skip to Content
Merck
All Photos(1)

Key Documents

18332

Millipore

Peptone from vegetable

suitable for microbiology

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41106212
NACRES:
NA.85
Pricing and availability is not currently available.

biological source

plant

Quality Level

form

powder

shelf life

limited shelf life, expiry date on the label

composition

amino-nitrogen, ≥3%
total nitrogen (N), ≥11%

impurities

≤15% ash

loss

≤6% loss on drying

pH

6.6±0.5 (25 °C, 2% in H2O)

solubility

H2O: 2%, clear, yellow

application(s)

food and beverages
vitamins, nutraceuticals, and natural products

microbiology

Looking for similar products? Visit Product Comparison Guide

General description

Peptone (vegetable) is a meat-free alternative to peptones from meat (Cat. No. 70164). It supports good growth of a wide variety of microorganisms in culture media in routine diagnostic and research bacteriology.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Debra Rossouw et al.
Applied and environmental microbiology, 75(20), 6600-6612 (2009-08-25)
Commercial wine yeast strains of the species Saccharomyces cerevisiae have been selected to satisfy many different, and sometimes highly specific, oenological requirements. As a consequence, more than 200 different strains with significantly diverging phenotypic traits are produced globally. This genetic

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service