Skip to Content
Merck
All Photos(2)

Key Documents

67386

Supelco

Sudan IV

analytical standard

Synonym(s):

Biebrich scarlet R fat soluble, Fat Ponceau R or 4, Lipid Crimson, Oil Red IV, Scarlet Red Scharlach, Solvent Red 24, Sudan R or BB

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H20N4O
CAS Number:
Molecular Weight:
380.44
Colour Index Number:
26105
Beilstein:
709018
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

Assay

≥96.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

199 °C (dec.) (lit.)

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

SMILES string

Cc1ccccc1\N=N\c2ccc(\N=N\c3c(O)ccc4ccccc34)c(C)c2

InChI

1S/C24H20N4O/c1-16-7-3-6-10-21(16)26-25-19-12-13-22(17(2)15-19)27-28-24-20-9-5-4-8-18(20)11-14-23(24)29/h3-15,29H,1-2H3/b26-25+,28-27+

InChI key

RCTGMCJBQGBLKT-PAMTUDGESA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1A

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yue Zhang et al.
Environmental toxicology, 26(3), 292-299 (2009-12-17)
We evaluated the role of oxidative stress in Sudan IV-induced DNA damage, using human liver-derived HepG2 cells. The DNA damaging effects of Sudan IV in HepG2 cells were evaluated by alkaline single cell gel electrophoresis assay and micronucleus test (MNT).
Quanlin Wang et al.
Se pu = Chinese journal of chromatography, 25(6), 864-866 (2008-02-09)
A reversed-phase high performance liquid chromatographic (RP-HPLC) method for the simultaneous determination of canthaxanthin, Sudan Red I , Sudan Red II , Sudan Red III and Sudan Red IV in salted duck egg yolk has been developed. These canthaxanthin and
Zhenxia Du et al.
Se pu = Chinese journal of chromatography, 25(5), 705-710 (2007-12-29)
A method of the simultaneous analysis of Sudan Red I - IV in duck egg yolk was developed with an ultra performance liquid chromatography-tandem mass spectrometry (UPLC-MS/MS). Acetonitrile was used to extract the residues from duck egg yolk, and water
Lichuan Niu et al.
Journal of the science of food and agriculture, 90(2), 338-342 (2010-04-01)
A green, simple and sensitive flow injection chemiluminescence (CL) procedure is proposed for the determination of Sudan dyes. The method is based on the finding that Sudan I, II, III and IV markedly enhance the CL intensity of the luminol/dissolved
Qi Wu et al.
Sheng wu gong cheng xue bao = Chinese journal of biotechnology, 25(10), 1477-1482 (2010-02-02)
We prepared the nano-sized magnetic fluid in aqueous phase solution to modify the killed yeast cells (Saccharomyces cerevisiae). Subsequently, the yeast cells with good magnetic response was obtained.The infrared spectroscopy analysis showed that the modified yeast cells increased obviously the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service