Skip to Content
Merck
All Photos(1)

Key Documents

64760

Sigma-Aldrich

4-Methoxyazobenzene

≥98.0%

Synonym(s):

4-Phenylazoanisole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H12N2O
CAS Number:
Molecular Weight:
212.25
Beilstein:
958054
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥98.0%

mp

54-56 °C

SMILES string

COc1ccc(cc1)\N=N\c2ccccc2

InChI

1S/C13H12N2O/c1-16-13-9-7-12(8-10-13)15-14-11-5-3-2-4-6-11/h2-10H,1H3/b15-14+

InChI key

LGCRPKOHRIXSEG-CCEZHUSRSA-N

Related Categories

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service