Skip to Content
Merck
All Photos(2)

Key Documents

S4261

Sigma-Aldrich

Sudan IV

Dye content, ≥80%, certified by the Biological Stain Commission, powder

Synonym(s):

Biebrich scarlet R fat soluble, Fat Ponceau R or 4, Lipid Crimson, Oil Red IV, Scarlet Red Scharlach, Solvent Red 24, Sudan R or BB

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H20N4O
CAS Number:
Molecular Weight:
380.44
Colour Index Number:
26105
Beilstein:
709018
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

Product Name

Sudan IV, certified by the Biological Stain Commission, BioXtra

grade

certified by the Biological Stain Commission

Quality Level

product line

BioXtra

form

powder

composition

Dye content, ≥80%

mp

199 °C (dec.) (lit.)

λmax

356-360 nm (2nd)
519-523 nm (1st)

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

Cc1ccccc1\N=N\c2ccc(\N=N\c3c(O)ccc4ccccc34)c(C)c2

InChI

1S/C24H20N4O/c1-16-7-3-6-10-21(16)26-25-19-12-13-22(17(2)15-19)27-28-24-20-9-5-4-8-18(20)11-14-23(24)29/h3-15,29H,1-2H3/b26-25+,28-27+

InChI key

RCTGMCJBQGBLKT-PAMTUDGESA-N

Looking for similar products? Visit Product Comparison Guide

Suitability

Certified as a fat staining using the Churukian variant of the Lillie-Ashburn supersaturated isopropanol method. Sudan IV is substituted for the oil red O in the procedure.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Sens. 1A

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Requirement of JNK1 for endothelial cell injury in atherogenesis
Narges Amini
Atherosclerosis (2014)
Hans F. Stroo
In Situ Remediation of Chlorinated Solvent Plumes (2010)
Histophatologic Techniques (2006)
The induction of cytochrome P450 1A1 by sudan dyes.
Refat NA
Journal of Biochemical and Molecular Toxicology (2008)
Jean T Jacob et al.
Investigative ophthalmology & visual science, 53(7), 3473-3480 (2012-04-19)
A lipophilic dye is used to investigate the degree to which the surface and bulk hydrophobic domains of the lenses can be imaged and to identify specific changes in the availability of those domains after in vitro wear and cleaning

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service